Skip to Content
Merck
All Photos(2)

Key Documents

P9290

Sigma-Aldrich

Proteinase K–Agarose from Tritirachium album

lyophilized powder

Synonym(s):

Proteinase K-Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

microbial (T.album
T. ALBUM)

Quality Level

form

lyophilized powder

extent of labeling

30-140 units per g solid

technique(s)

DNA extraction: suitable
RNA extraction: suitable
protein extraction: suitable

swelling

1 g swells to ~5 mL

storage temp.

−20°C

Application

Proteinase K-agarose is used in protein and nucleic acid isolation, modifying enzymes and immobilized enzymes. Proteinase K-agarose has been used to study Klebsiella pneumonia as well as Echinococcus multilocularis infection in domesticated dogs.

Biochem/physiol Actions

The application of proteinase K has been extended by immobilizing it on an insoluble matrix such as agarose. The agarose is quite inert in most systems and allows high activities per unit weight. This reactive resin allows the enzyme reaction to be quickly catalyzed by momentary contact with the substrate in a suitable medium. Reactions can be carried out in a batch slurry or even through a small column. In either case, the insoluble enzyme does not contaminate the reaction mix, which can be readily filtered out of the batch slurry or remain behind in the column.

Features and Benefits

• Recylable Proteinase K on matrix support.
• Spin column or batch processing possible.

Unit Definition

One unit will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent).

Physical form

Lyophilized powder stabilized with lactose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shai Eyal et al.
Development (Cambridge, England), 146(14) (2019-06-22)
Bone protrusions provide stable anchoring sites for ligaments and tendons and define the unique morphology of each long bone. Despite their importance, the mechanism by which superstructures are patterned is unknown. Here, we identify components of the genetic program that
Yuan Gao et al.
Zhonghua liu xing bing xue za zhi = Zhonghua liuxingbingxue zazhi, 31(7), 786-790 (2010-12-18)
To type Klebsiella pneumonia through methods including pulse-field gel electrophoresis (PFGE) in combination with multilocus sequence typing. Four selected different EPs, referring to the Standard Operating Procedure of PulseNet China, were used. The single colony of Klebsiella pneumonia was quantified
Guillaume Saubeau et al.
Molecules (Basel, Switzerland), 19(10), 15374-15390 (2014-09-30)
The induction of plant immunity by Pathogen Associated Molecular Patterns (PAMPs) constitutes a powerful strategy for crop protection. PAMPs indeed induce general defense responses in plants and thus increase plant resistance to pathogens. Phytophthora infestans culture filtrates (CCFs) are known
M J Pearse et al.
Immunology letters, 18(3), 219-223 (1988-07-01)
This report describes a method for preparing both DNA and RNA simultaneously from as few as 5 X 10(5) lymphoid cells. The method is suitable for cultured cells or any tissue from which a cell suspension can be prepared and
Shai Eyal et al.
Development (Cambridge, England), 146(4) (2019-02-13)
Sesamoid bones are small auxiliary bones that form near joints and contribute to their stability and function. Thus far, providing a comprehensive developmental model or classification system for this highly diverse group of bones has been challenging. Here, we compare

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service