Skip to Content
Merck

P8563

Polyuridylic acid–Agarose

lyophilized powder, matrix polyacrylhydrazido-agarose

Synonym(s):

Poly(U)–Agarose potassium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Segment

extent of labeling

0.1-1.0 mg per mL

matrix

polyacrylhydrazido-agarose

matrix attachment

ribose moiety after periodate oxidation

swelling

1 g swells to ~4 mL

capacity

≥0.2 mg/mL binding capacity (poly(A) (P 9403))

storage temp.

−20°C

Application

Polyuridylic acid-agarose is used in protein chromatography, affinity chromatography and specialty resins. Polyuridylic acid has been used to study insulin receptors and to develop polyclonal antibodies against RNase L.

Physical form

Lyophilized powder stabilized with lactose


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library