Skip to Content
Merck
All Photos(1)

Key Documents

P8115

Sigma-Aldrich

Extract-N-Amp PCR ReadyMix for Blood

12 mL sufficient for 1000 amplifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

Quality Level

usage

sufficient for 1000 reactions
12 mL sufficient for 1000 amplifications

feature

dNTPs included
hotstart

color

colorless

shipped in

wet ice

storage temp.

−20°C

General description

The Extract-N-Amp Blood PCR Kits contain all the reagents needed to rapidly extract and amplify human genomic DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

Application

Extract-N-Amp PCR ReadyMix for Blood has been used in the polymerase chain reaction (PCR).

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
GenElute is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Götz Münchow et al.
Expert review of molecular diagnostics, 5(4), 613-620 (2005-07-15)
A chip-based PCR device is presented that is capable of rapid temperature ramping and handling sample volumes in the microliter range. The PCR chip comprises a microchannel thermally connected to three temperature zones. Inside this microchannel, the PCR sample plug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service