P6366
Nunc® MaxiSorp™ 384 well plates
384 well plate, clear, polystyrene, 120ul/well, pinchbar design, 30/cs
Synonym(s):
384 multiwell plate, 384 well microplate, 384 well microtiter plate, 384 well plate, ELISA plate, immunoassay plate
About This Item
flat bottom clear polystyrene wells
Recommended Products
material
clear polystyrene
flat bottom clear polystyrene wells
sterility
non-sterile
feature
lid: no
packaging
case of 30 ea (internal packs of 10)
manufacturer/tradename
Nunc 464718
maximum volume
120 μL
plate L × W
128 mm × 86 mm
size
384 wells
well maximum volume
120 μm
working volume
100 μL
suitability
suitable for (colorimetric assays, sample storage; luminescence and fluorescence assays, ELISA)
binding type
(MaxiSorp surface)
Looking for similar products? Visit Product Comparison Guide
General description
- Same footprint as a 96 well plate
- Working volume range of 10-100 μL
- Compatible with robotic, automated readers and liquid handling systems
- Assay miniaturization increases productivity and reduces cost
- C-well for optical clarity and sample recovery
- Reduced risk of well-to-well contamination
- Optimizes robotic access to wells
- White and black plates minimize crosstalk
- Rounded square wells eliminate wicking (capillary action)
- Flat bottom for optical reading
- Optimized for scintillation counting
- Black 384 well plate reduces background fluorescence and minimizes backlight scatter
- White 384 well plate maximizes reflection of signal while maintaining low background
- Clear 384 well plate for optical clarity for readthrough applications
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service