Skip to Content
Merck
All Photos(3)

Key Documents

P5542

Sigma-Aldrich

Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus

lyophilized powder, ≥1,000 units/mg protein (Lowry)

Synonym(s):

1-Phosphatidyl-D-myo-inositol inositol phosphohydrolase, cyclic-phosphate forming, PI-PLC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

lyophilized powder

Quality Level

specific activity

≥1,000 units/mg protein (Lowry)

foreign activity

Phopholipase C (lecithinase) ≤1 units/mg protein
Sphingomyelinase ≤40 units/mg protein

storage temp.

−20°C

Gene Information

Bacillus cereus E33L ... BCZK3513(3026815)

Looking for similar products? Visit Product Comparison Guide

General description

Phosphatidylinositol-specific phospholipase C (PI-PLC) from Bacillus cereus is a 298 residual protein. It belongs to the class of ubiquitous enzymes. PI-PLC is characterized with a single globular domain and it folds into eight (βα) barrel sheets, out of which only six βα units are found in barrel, lacking helices between strands IV and V and V and VI.

Application

Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus has been used to cleave GPI-anchored protein from green fluorescent protein- glycosylphosphatidylinositol (GFP-GPI) from the cell membrane. It has also been used in invasion assay to test the involvement of PLAP in T. cruzi invasion to human epithelial type 2 (HEp-2) cells.

Biochem/physiol Actions

Used for the release of GPI anchored proteins from the membrane.

Unit Definition

One unit will liberate one unit of acetylcholinesterase per minute from a membrane-bound crude preparation at pH 7.4 at 30 °C (10 minute incubation).

Physical form

Contains phosphate buffer salts, EDTA and stabilizer

Analysis Note

Acetylcholinesterase is measured according to Ellman, et al.

inhibitor

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of the phosphatidylinositol-specific phospholipase C from Bacillus cereus in complex with myo-inositol.
Heinz DW
The Embo Journal, 14(16), 3855-3863 (1995)
Danielle K Bailey et al.
PloS one, 16(7), e0254794-e0254794 (2021-07-27)
Iron and other transition metals, such as copper and manganese, are essential for supporting brain function, yet over-accumulation is cytotoxic. This over-accumulation of metals, particularly iron, is common to several neurological disorders; these include Alzheimer's disease, Parkinson's disease, Friedrich's ataxia
Role of placental alkaline phosphatase in the internalization of trypomatigotes of Trypanosoma cruzi into HEp2 cells.
Sartori MJ
Tropical Medicine & International Health, 8(9), 832-839 (2003)
Imaging of mobile long-lived nanoplatforms in the live cell plasma membrane.
Brameshuber M
The Journal of Biological Chemistry, 285(53), 41765-41771 (2010)
Helen Neuert et al.
Development (Cambridge, England), 147(2) (2019-12-22)
The development of tissues and organs requires close interaction of cells. To achieve this, cells express adhesion proteins such as the neural cell adhesion molecule (NCAM) or its Drosophila ortholog Fasciclin 2 (Fas2). Both are members of the Ig-domain superfamily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service