Skip to Content
Merck
All Photos(2)

Key Documents

P4109

Sigma-Aldrich

β-Phosphoglucomutase from Lactococcus sp.

recombinant, expressed in E. coli

Synonym(s):

β-D-Glucose 1,6-phosphomutase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

form

solid

specific activity

≥10 unit/mg solid

storage temp.

−20°C

Related Categories

Application

β-Phosphoglucomutase is used to study starch and sucrose metabolism[1]. It is used for the determination of α-amylase in clinical analysis. It is used to study galactose utilization[2] and generalized glycogenosis (Pompe′s Disease)[3].

Biochem/physiol Actions

β-Phosphoglucomutase enzymatically converts β-D-glucose 1-phosphate to β-D-glucose 6-phosphate. It is involved in starch and sucrose metabolism[1].
Enzymatically converts β-D-glucose-1-phosphate to β-D-glucose-6-phosphate.

Unit Definition

One unit is defined as the amount of enzyme which converts 1μmol of β-D-glucose-1-phosphate to β-D-glucose-6-phosphate per minute at 37°C and pH 7.0.

Other Notes

Contains EDTA and lactose.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brigitta Elsässer et al.
Journal of molecular modeling, 18(7), 3169-3179 (2012-01-13)
Enzyme catalyzed phosphate transfer is a part of almost all metabolic processes. Such reactions are of central importance for the energy balance in all organisms and play important roles in cellular control at all levels. Mutases transfer a phosphoryl group
Cecelia A Garcia et al.
Applied microbiology and biotechnology, 105(10), 4033-4052 (2021-05-08)
In a competitive microbial environment, nutrient acquisition is a major contributor to the survival of any individual bacterial species, and the ability to access uncommon energy sources can provide a fitness advantage. One set of soluble carbohydrates that have attracted
OCULAR HISTOPATHOLOGY IN GENERALIZED GLYCOGENOSIS (POMEPE'S DISEASE).
D TOUSSAINT et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 73, 342-349 (1965-03-01)
F Levander et al.
Applied and environmental microbiology, 67(6), 2734-2738 (2001-05-26)
To study the influence of phosphoglucomutase (PGM) activity on exopolysaccharide (EPS) synthesis in glucose- and lactose-growing Streptococcus thermophilus, a knockout PGM mutant and a strain with elevated PGM activity were constructed. The pgmA gene, encoding PGM in S. thermophilus LY03
Erika Pellegrini et al.
Acta crystallographica. Section D, Biological crystallography, 67(Pt 10), 902-906 (2011-09-21)
Over the last 20 years cryocrystallography has revolutionized the field of macromolecular crystallography, greatly reducing radiation damage and allowing the collection of complete data sets at synchrotron sources. However, in order to cool crystals to 100 K cryoprotective agents must usually

Questions

  1. Hello, I received the enzyme pgmB (P4109) and wondered in what buffer should I dissolve the enzyme powder. Thank you

    1 answer
    1. This product may be reconstituted in 10 mM Potassium Phosphate, pH 7 at 5mg/mL. The stock solution is expected to be stabe for at least 1 week when stored refrigerated. For assay purposes, the enzyme is prepared just prior to use with ice cold 10 mM Potassium Phosphate, pH 7 solution duluted to 1.0 - 3.0 U/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service