Skip to Content
Merck
All Photos(3)

Key Documents

P3300

Sigma-Aldrich

Monoclonal Anti-Phosphotyrosine antibody produced in mouse

clone PT-66, ascites fluid

Synonym(s):

Monoclonal Anti-Phosphotyrosine, Phospho-Tyr, Phospho-tyrosine, p-Tyr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PT-66, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:1,600
western blot: 1:2,000 using a fresh human platelet preparation

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

As determined by ELISA and competitive ELISA, the antibody reacts specifically with phosphorylated tyrosine, both as free amino acid or conjugated to carriers such as BSA or KLH. No cross-reactivity is observed with non-phosphorylated tyrosine, phosphothreonine, phosphoserine, AMP or ATP.
Monoclonal Anti-Phosphotyrosine (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
Phosphorylation, attachment of a phosphate group to protein, alters protein functionality by activation or deactivation of the protein, in which protein kinase plays a crucial role. Tyrosine residue phosphorylation plays a key role in cell growth and differentiation. The monoclonal anti-phosphotyrosine antibody is useful in immunoblotting for identification of phosphotyrosine containing protein from cultured human epidermoid carcinoma cell line A-431 and human platelets preparation. This product can be used in immunofluorescent labeling of tyrosine residue at focal adhesion and cellular junctions of cultured MDCK cells. This product is also useful in studies of signal transduction and growth factor receptors. This antibody has shown specificity for binding to proteins that contain phosphorylated tyrosine residue either in free amino acid form or in conjugation with KLH or BSA but will not react with non-phosphorylated tyrosine or other phosphorylated proteins and amino acids. It will also not react with phosphorylated molecules like ATP or AMP. Monoclonal anti-phosphotyrosine reacts specifically with mouse and human.

Specificity

This antibody is specific for phosphorylated tyrosine both as the free amino acid or when conjugated to carriers such as BSA or KLH

Immunogen

phosphotyrosine conjugated to BSA

Application

Monoclonal Anti-Phosphotyrosine antibody produced in mouse has been used in
  • immunocytochemistry
  • immunoprecipitation
  • western blotting
  • peptide binding studies flow cytometry
  • enzyme linked immunosorbent assay (ELISA)
  • radioimmunoassay (RIA)
  • immunoaffinity isolation

Monoclonal anti-phosphotyrosine antibody produced in mouse is used in immunohistochemisty, flow cytometry, immunoprecipitation, immunoblotting, ELISA and RIA for localization of phosphorylated tyrosine containing proteins. It can also be used for immunoaffinity isolation.

Biochem/physiol Actions

Phosphotyrosine levels enhanced in cellular proteins leads to activation of several cellular processes mediated by phosphotyrosine kinases. Phosphotyrosine residues that are autophosphorylated by the binding of ligands associated with receptors like epidermal growth factor (EGF), platelet-derived growth factor (PDGF) and insulin receptors, functions as the primary integral component of mitogenic signaling cascade. Phosphotyrosine mediates the activation of T-cell as a result of phosphorylation of tyrosine residues by tyrosine kinases in cytoplasmic domains of CD4 an CD8 which in turn phosphorylates TCR-CD3 complex. Phosphotyrosine activity has also been found in many retroviruses whose oncogenes encodes tyrosine-specific protein kinase.
Tyrosine residue phosphorylation plays a key role in cell growth and differentiation.

Physical form

This product is supplied as ascites fluid containing 15 mM sodium azide

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyrosine kinase-independent activation of extracellular-regulated kinase (ERK) 1/2 by the insulin-like growth factor-1 receptor
Perrault R, et al.
Cellular Signalling, 23(4), 739-746 (2011)
Katrin Schmelzle et al.
Diabetes, 55(8), 2171-2179 (2006-07-29)
The insulin-signaling network regulates blood glucose levels, controls metabolism, and when dysregulated, may lead to the development of type 2 diabetes. Although the role of tyrosine phosphorylation in this network is clear, only a limited number of insulin-induced tyrosine phosphorylation
Neuropathology in mice expressing human alpha-synuclein
van der Putten H, et al.
The Journal of Neuroscience, 20(16), 6021-6029 (2000)
S Barlati et al.
FEBS letters, 281(1-2), 137-140 (1991-04-09)
Immunoblotting analysis of purified human urokinase plasminogen activator (u-PA), gives a positive signal when reacted with anti-phosphotyrosine monoclonal antibodies (MoAb anti-P-Tyr); competition with o-phospho-DL-tyrosine (P-Tyr) but not o-phospho-DL-threonine or serine (P-Treo, P-Ser) completely suppresses this signal. Either the 55 kDa
Epidermal Growth Factor Receptor Down-Regulation Triggers Human Myoblast Differentiation
Boisguerin P, et al.
Chembiochem, 8(18), 2302-2307 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service