Skip to Content
Merck
All Photos(2)

Documents

P3292

Sigma-Aldrich

Pancreatin from porcine pancreas

powder, suitable for cell culture, 4 × USP specifications

Synonym(s):

Pancreatin from hog pancreas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75

biological source

Porcine pancreas

Agency

USP (specifications)

form

powder

specific activity

4 × USP specifications

contains

lactose or sucrose as extender (The sucrose used might contain up to 3.25% starch.)

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Pancreatin from porcine pancreas has been used to assess the treatment of steatorrhea by lipase supplementation therapy, to investigate treatment options for pancreatic diabetes in patients experiencing the decompensated stage of chronic pancreatitis, and to safely and effectively remove formalin-fixed tissues from arterial grafts without leading to structural damage or loss in fiber integrity.

Biochem/physiol Actions

Pancreatin contains enzymatic components including trypsin, amylase and lipase, ribonuclease, and protease, produced by the exocrine cells of the porcine pancreas. This combination of enzymes allows it to hydrolyze proteins, starch and fats. Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40°C, will digest not less than 25 times its weight of casein in 60 minutes at pH 7.5 at 40°C and will release not less than microequivalents of acid per min per mg pancreatin from olive oil at pH 9.0 at 37°C.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel N Meijles et al.
The Biochemical journal, 478(11), 2059-2079 (2021-05-19)
Insulin and insulin-like growth factor stimulate protein synthesis and cardioprotection in the heart, acting through their receptors (INSRs, IGF1Rs) and signalling via protein kinase B (PKB, also known as Akt). Protein synthesis is increased in hearts perfused at alkaline pHo
Xu-Dong Kong et al.
Nature biomedical engineering, 4(5), 560-571 (2020-05-13)
The oral administration of peptide drugs is hampered by their metabolic instability and limited intestinal uptake. Here, we describe a method for the generation of small target-specific peptides (less than 1,600 Da in size) that resist gastrointestinal proteases. By using
Hyunji Byun et al.
Reproductive biology and endocrinology : RB&E, 19(1), 112-112 (2021-07-18)
The tumor susceptibility gene 101 (Tsg101), a component of the endosomal sorting complex required for transport (ESCRT) complex I, is involved in multiple biological processes involving endomembranous structures and the plasma membrane. The role of Tsg101 in the uterine epithelium
C Giromini et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(8), 1436-1445 (2017-04-12)
Ex-food or former food products (FFPs) have been proposed as one of the categories with great promise as alternative feed ingredients. FFPs' nutritional potential is not yet fully exploited. Therefore, the aim of this study was to perform a nutritional
Luisina Ongaro et al.
Biology of reproduction, 102(3), 639-646 (2019-11-15)
Activins selectively stimulate follicle-stimulating hormone (FSH) secretion by pituitary gonadotrope cells. More recently, other members of the TGFbeta superfamily, the bone morphogenetic proteins (BMPs), were reported to regulate FSH synthesis. Activins and BMPs independently and synergistically stimulate transcription of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service