P2200
Protein C from human plasma
Activated, lyophilized powder, ≥90% (SDS-PAGE)
Synonym(s):
Activated Protein C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human plasma
Quality Level
Assay
≥90% (SDS-PAGE)
form
lyophilized powder
mol wt
heavy chain 41 kDa
light chain 21 kDa
technique(s)
inhibition assay: suitable
solubility
H2O: 1 mg/mL
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... PROC(5624)
General description
Protein C from human plasma is encoded by the gene PROC. In human chromosome, the gene is localised on chromosome 2q14. Activated protein C (APC) cleaves protease activated receptor 1 (PAR1) resulting in cytoprotective and anti-inflammatory effects. Clinical trials with APC implicates its use in treating severe early onset preeclampsia in pregnant women and prolongs pregnancy and helps in perinatal outcomes.
Application
Protein C from human plasma has been used for pre-treatment of endothelial cells prior to antibody-inhibition assay. It has also been used in activated protein C (APC) assay to determine its inhibitory effect on copper.
Biochem/physiol Actions
In addition, activated protein C has been shown to inhibit TNF-α induced expression of the inflammatory proteins VCAM, ICAM-I, and Ilk-8 in endothelial cells.
Protein C is a plasma, vitamin κ-dependent zymogen of a serine protease that can inhibit blood coagulation by inhibiting thrombin formation, selectively inactivating Factors Va and VIIIa.
Protein C is a plasma, vitamin κ-dependent zymogen of a serine protease that can inhibit blood coagulation by inhibiting thrombin formation, selectively inactivating Factors Va and VIIIa. The Protein C anticoagulant pathway is triggered when thrombin binds to the endothelial cell proteoglycan, thrombomodulin. This complex, which cannot clot blood, is a potent activator of the protein C zymogen. Activation involves the release of a dodecapeptide from the N-terminal domain of the heavy chain. The activated Protein C (APC) then binds to protein S on cell surfaces and inactivates the coagulation factors Va and VIIIa by proteolysis. APC has also been shown to bind to receptors on the endothelium of large blood vessels.
Physical form
Lyophilized powder from 20 mM Tris-HCl, pH 7.4, containing 0.1 M NaCl
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Journal of biological chemistry, 261(32), 14991-14996 (1986-11-15)
The kinetic properties of the activation by monovalent cations of the amidolytic activity of bovine des-1-41 light chain activated protein C have been examined. With the cations Cs+, K+, Li+, and Tl+, a single cation site, or class of sites
Dose-response : a publication of International Hormesis Society, 18(2), 1559325820917288-1559325820917288 (2020-05-20)
Although the effect of activated protein C (APC) on neuronal injury and neuroinflammatory responses has been extensively studied, the detailed mechanism underlying APC-protective effect in the blood-brain barrier (BBB) injury during ischemia is still not clear. In this study, the
Nature communications, 13(1), 1222-1222 (2022-03-11)
Many individual genetic risk loci have been associated with multiple common human diseases. However, the molecular basis of this pleiotropy often remains unclear. We present an integrative approach to reveal the molecular mechanism underlying the PROCR locus, associated with lower
The Korean journal of pain, 32(3), 168-177 (2019-07-02)
Brennan's rodent paw incision model has been extensively used for understanding mechanisms underlying postoperative pain in humans. However, alterations of physiological parameters like blood pressure and heart rate, or even feeding and drinking patterns after the incision have not been
Severe malaria is associated with parasite binding to endothelial protein C receptor
Nature, 498(7455), 502-502 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service