Skip to Content
Merck
All Photos(1)

Documents

P1925

Millipore

Protein A–Agarose macrobeads

aqueous suspension

Synonym(s):

Protein A resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

aqueous suspension

Quality Level

extent of labeling

~6 mg per mL

matrix activation

cyanogen bromide

matrix spacer

1 atom

capacity

~6 mg/mL binding capacity (human IgG)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Protein A is an immunoglobulin (Ig)-binding protein that might be suitable for low-pressure antibody isolation. It is used to purify large amounts of IgG and binds to the Fc part of the antibody at the CH2–CH3 interface. Protein A-agarose is used for affinity chromatography, antibody purification and characterization, and protein A, G and L resins.

Application

Protein A–Agarose macrobeads has been used in immunoprecipitation. It has been used to study the effects of protein A immunoadsorption in patients with chronic dilated cardiomyopathy as well as to study multiple sclerosis and gastric cancer.

Physical form

Suspension in 0.9% NaCl containing 15Mm Sodium Azide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Blumbach et al.
Journal of cell science, 111 ( Pt 17), 2635-2644 (1998-08-14)
Porifera (sponges) are the oldest extant metazoan phylum. Dissociated sponge cells serve as a classic system to study processes of cell reaggregation. The reaggregation of dissociated cells is mediated by an extracellularly localized aggregation factor (AF), based on heterophilic interactions
Human IgG Subclasses
Antibody Fc, 159-177 (2014)
C Wagner-Hülsmann et al.
Glycobiology, 6(8), 785-793 (1996-12-01)
The cDNA for the full-length lectin from the marine sponge Geodia cydonium was cloned. Analysis of the deduced aa sequence revealed that this lectin belongs to the group of galectins. The full-length galectin, which was obtained also in a recombinant
C Cullin et al.
Yeast (Chichester, England), 10(1), 105-112 (1994-01-01)
In this paper are described a set of new high-copy-number yeast vectors, which are specially designed for the conditional expression of epitope-tagged proteins in vivo. One of the major advantages of these plasmids is that they allow polymerase chain reaction-amplified
Nanmei Liu et al.
The Journal of nutritional biochemistry, 18(9), 597-608 (2007-03-17)
Transcuprein is a high-affinity copper carrier in the plasma that is involved in the initial distribution of copper entering the blood from the digestive tract. To identify and obtain cDNA for this protein, it was purified from rat plasma by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service