Skip to Content
Merck
All Photos(2)

Key Documents

P1537

Sigma-Aldrich

Polyadenylic acid-Polyuridylic acid sodium salt

double-stranded homopolymer

Synonym(s):

Poly[A]•Poly[U]

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

Quality Level

form

powder

storage temp.

−20°C

SMILES string

[P](=O)(OC[C@H]3O[C@H]([C@@H]([C@@H]3O)O)[n]4c5ncnc(c5nc4)N)(O)O.[P](=O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)N2C=CC(=O)NC2=O)(O)O

InChI

1S/C10H14N5O7P.C9H13N2O9P/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(22-10)1-21-23(18,19)20;12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20);1-2,4,6-8,13-14H,3H2,(H,10,12,15)(H2,16,17,18)/t4-,6-,7-,10-;4-,6-,7-,8-/m11/s1

InChI key

VGQHQOKIMNKUEF-ZLOOHWKQSA-N

Application

Polyadenylic acid-Polyuridylic acid (polyAU) is a double stranded homodimer studied for potential therapeutic activity, especially when coupled with radiotherapy. PolyA/U is used in comparative physiochemical studies with other duplex and single-stranded polymers such as poly-A and poly-U.

Other Notes

Double stranded homopolymer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Laplanche et al.
Breast cancer research and treatment, 64(2), 189-191 (2001-02-24)
With a median follow-up of 14 years, the combination of polyadenylic-polyuridylic acid plus locoregional radiotherapy (257 patients) has significantly improved disease-free survival (p = 0.03) and significantly reduced the incidence of metastases (p = 0.04) when compared to CMF alone
Polyadenylic acid-polyuridylic acid (poly A : U) and experimental murine brucellosis. II. Macrophages as target cells of poly A : U in experimental brucellosis.
E D Madraso et al.
Immunology, 35(1), 77-84 (1978-07-01)
Rosa Conforti et al.
Cancer research, 70(2), 490-500 (2010-01-14)
Many cancer cells express Toll-like receptors (TLR) that offer possible therapeutic targets. Polyadenylic-polyuridylic acid [poly(A:U)] is an agonist of the Toll-like receptor TLR3 that displays anticancer properties. In this study, we illustrate how the immunostimulatory and immunosuppressive effects of this
H-C Jeung et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 19(3), 520-526 (2007-11-22)
This phase III trial was to compare 5-fluorouracil (5-FU), adriamycin, and polyadenylic-polyuridylic acid (poly A:U) against 5-fluorouracil plus adriamycin (FA) for operable gastric cancer. From 1984 to 1989, patients who had D(2-3) curative resection were randomly assigned to receive chemotherapy
Nino Lomadze et al.
Organic & biomolecular chemistry, 4(9), 1755-1759 (2006-04-25)
The affinities of polyamines consisting of ethylenediamine units equipped with either one or two terminal naphthyl-, anthryl-, or acridyl units towards PolyA.PolyU as an RNA model, and Poly(dA).Poly(dT) as a DNA model are screened by measuring the melting point changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service