Skip to Content
Merck
All Photos(2)

Key Documents

P0054

Sigma-Aldrich

Anti-PIP5K3 (N-terminal) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-1-phosphatidylinositol-4-phosphate 5-kinase, Anti-Fab1, Anti-Fyve finger-containing phosphoinositide kinase, Anti-P235, Anti-PIKfyve, Anti-PIP5K, Anti-Phosphatidylinositol-3-phosphate/phosphatidylinositol 5-kinase, type III

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 2-4 μg/mL using HEK-293T cells expressing human PIP5K3.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Phosphatidylinositol-3-phosphate-5-kinase, type 3 (PIP5K3, also known as PIKfyve and FAB1) belongs to a large family of lipid kinases that modify the phosphorylation state of phosphatidylinositol (PtdIns). PIP5K3 has been shown to be localized on early and late endosomes.
Phosphatidylinositol-3-phosphate/phosphatidylinositol-5-kinase, type III (PIP5K3) is a constituent of a trimolecular protein complex. It is a 2089 amino acid protein. The gene encoding it is located on human chromosome 2 and has 41 exons.

Specificity

Anti-PIP5K3 (N-terminal) specifically recognizes human PIP5K3.

Application

Anti-PIP5K3 (N-terminal) antibody produced in rabbit can be used in several immunochemical techniques including immunoblotting.

Biochem/physiol Actions

Phosphatidylinositol-3-phosphate-5-kinase, type 3 (PIP5K3) enzymatic activity is critical for cell morphology and endocytic membrane homeostasis. PIP5K3 acts as a negative regulator of endosome function or regulating endosome-trans-golgi-network (TGN) trafficking. It negatively regulates exocytosis on neurosecretory cells. PIP5K3 regulates the trafficking of IRAP/GLUT4 vesicles to plasma membrane in adipocytes, suggesting that it may play a pleiotropic role in membrane trafficking, including the exocytic pathway.
Phosphatidylinositol-3-phosphate/phosphatidylinositol-5-kinase, type III (PIP5K3) is involved in the synthesis of phosphatidylinositol 3, 5-bisphosphate. It also possesses a protein kinase activity. PIP5K3 regulates specific signaling pathways as well as membrane trafficking. Mutations in the gene encoding it are associated with fleck corneal dystrophy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion of PIKfyve alters alveolar macrophage populations and exacerbates allergic inflammation in mice
Kawasaki T, et al.
The Embo Journal, 36(12), 1707-1718 (2017)
Protein kinase B phosphorylation of PIKfyve regulates the trafficking of GLUT4 vesicles
Berwick DC, et al.
Journal of Cell Science, 117(25), 5985-5993 (2004)
A novel PIKFYVE mutation in fleck corneal dystrophy
Andreas K
Molecular Vision (2011)
Functional dissociation between PIKfyve-synthesized PtdIns5P and PtdIns(3,5)P2 by means of the PIKfyve inhibitor YM201636
Diego Sbissa
American Journal of Physiology. Cell Physiology (2012)
Localized PtdIns 3, 5-P2 synthesis to regulate early endosome dynamics and fusion
Ikonomov OC, et al.
The American Journal of Physiology, 291(2), C393-C404 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service