Skip to Content
Merck
All Photos(1)

Key Documents

O9264

Sigma-Aldrich

N-(3-Oxotetradecanoyl)-L-homoserine lactone

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31NO4
Molecular Weight:
325.44
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

N-(3-Oxotetradecanoyl)-L-homoserine lactone,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white

application(s)

cell analysis

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)CC(=O)N[C@H]1CCOC1=O

InChI

1S/C18H31NO4/c1-2-3-4-5-6-7-8-9-10-11-15(20)14-17(21)19-16-12-13-23-18(16)22/h16H,2-14H2,1H3,(H,19,21)/t16-/m0/s1

InChI key

YQFJJDSGBAAUPW-INIZCTEOSA-N

Application

N-(3-Oxotetradecanoyl)-L-homoserine lactone (3-Oxo-C14-HSL), a member of a family of acyl homoserine lactones, may be used to determine its mechanisms and specificity of action as a signaling molecule involved in the regulation of bacterial quorum sensing.

Biochem/physiol Actions

Acyl homoserine lactone used as an autoinducer of quorum signaling by Sinorhizobium meliloti, Yersinia enterocolitica, and other Gram-negative bacteria.
N-(3-Oxotetradecanoyl)-L-homoserine lactone (3-oxo-C14-HSL) is among a group of homoserine lactones that includes N-octanoyl-homoserine lactone (N-C8-HSL), N-(3-oxodecanoyl) homoserine-L-lactone (3-oxo-C10 HSL), N-(3-oxododecanoyl)homoserine-L-lactone (3-oxo-C12-HSL), N-(3-hydroxydecanoyl)-L-homoserine lactone, and N-(3-hydroxyoctanoyl)-L-homoserine lactone involved in the processes of bacterial quorum sensing. These homoserine lactones are used to study the processes and mechanisms of bacterial quorum sensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mair E A Churchill et al.
Methods in molecular biology (Clifton, N.J.), 692, 159-171 (2010-10-30)
Quorum sensing plays a central role in regulating many community-derived symbiotic and pathogenic relationships of bacteria, and as such has attracted much attention in recent years. Acyl-homoserine lactones (AHLs) are important signaling molecules in the quorum sensing gene-regulatory processes found
Manabu Horikawa et al.
Bioorganic & medicinal chemistry letters, 16(8), 2130-2133 (2006-02-08)
The synthesis of the analogs of N-3-oxododecanoyl-L-homoserine lactone (1) and their structure-activity relationship for the apoptotic induction in macrophages, P388D1 cells, are described. It was revealed that the position of the oxo group in the acyl side chain in addition
Rumana Rashid et al.
Microbes and environments, 26(2), 144-148 (2011-04-20)
N-Acylhomoserine lactones (AHLs) are used as quorum-sensing signaling molecules by many Gram-negative bacteria. Here, 413 bacterial strains were obtained from the roots of potato plants and screened for AHL-degrading bacteria using Chromobacterium violaceum reporter strains. Sixty one isolates degraded N-hexanoyl-L-homoserine
John F Teiber et al.
Methods in molecular biology (Clifton, N.J.), 692, 291-298 (2010-10-30)
Mammalian paraoxonases (PONs) are a unique, highly conserved family of calcium-dependent esterases consisting of PON1, PON2, and PON3. The PONs can hydrolyze the lactone ring of a range of N-acyl-L: -homoserine lactone (AHL) quorum sensing signaling molecules, rendering them inactive.
Leo Eberl
International journal of medical microbiology : IJMM, 296(2-3), 103-110 (2006-02-24)
The genus Burkholderia contains over 30 species, many of which are important human pathogens. In addition to the primary pathogens Burkholderia pseudomallei and Burkholderia mallei, several species have emerged as opportunistic pathogens in persons suffering from cystic fibrosis (CF) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service