Skip to Content
Merck
All Photos(1)

Documents

O6887

Sigma-Aldrich

[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-Me-Tyr2, Orn8]-Oxytocin

≥93% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H74N12O12S2
CAS Number:
Molecular Weight:
1075.30
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥93% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC[C@H](C)C1NC(=O)[C@H](Cc2ccc(OC)cc2)NC(=O)CC3(CCCCC3)SSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(N)=O)NC1=O)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CCCN)C(=O)NCC(N)=O

InChI

1S/C48H74N12O12S2/c1-4-27(2)40-46(70)56-31(16-17-36(50)61)42(66)57-33(23-37(51)62)43(67)58-34(47(71)60-21-9-11-35(60)45(69)55-30(10-8-20-49)41(65)53-25-38(52)63)26-73-74-48(18-6-5-7-19-48)24-39(64)54-32(44(68)59-40)22-28-12-14-29(72-3)15-13-28/h12-15,27,30-35,40H,4-11,16-26,49H2,1-3H3,(H2,50,61)(H2,51,62)(H2,52,63)(H,53,65)(H,54,64)(H,55,69)(H,56,70)(H,57,66)(H,58,67)(H,59,68)/t27-,30-,31-,32-,33-,34-,35-,40-/m0/s1

InChI key

ZWVZXPFUQHTUKV-SZQHDTAUSA-N

Gene Information

Amino Acid Sequence

β-Mercapto-β,β-Cyclopentamethylenepropionyl-MeTyr-Ile-Gln-Asn-Cys-Pro-Orn-Gly-NH2 [Disulfide Bridge: 1-6]

Application

[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-Me-Tyr2, Orn8]-Oxytocin has been used as an oxytocin receptor antagonist:
  • to study the influence of blockading the oxytocin receptors in the anterior cingulate cortex on helping behavior
  • to determine the receptors that mediate vasopressin-stimulated anion secretion across cultured porcine vas deferens epithelial cell monolayers
  • to study the influence of oxytocin receptors inhibition on intravenous and in situ oxytocin effects on the urinary bladder and cardiovascular activities in sham rats

Biochem/physiol Actions

Potent oxytocin antagonist.
[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-Me-Tyr2, Orn8]-also acts as an arginine vasopressin receptor 1A (AVPR1A) antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Lambert et al.
Neuroscience, 57(4), 1027-1038 (1993-12-01)
During suckling, the periodic and synchronous bursting activity of oxytocin neurons has been shown to be facilitated by oxytocin itself, acting via several target sites, including the magnocellular nuclei. To investigate the role of the endogenous oxytocin released within the
R J Windle et al.
The Journal of endocrinology, 152(2), 257-264 (1997-02-01)
The effect of three oxytocin receptor antagonists on the renal actions of oxytocin and vasopressin was investigated in conscious male rats infused with hypotonic saline. Infusion of oxytocin at 100 pg/min produced plasma concentrations of 12.7 +/- 3.3 pmol/l and
M R Melis et al.
European journal of pharmacology, 294(1), 1-9 (1995-12-27)
In order to provide further support for a role of central nitric oxide as a mediator of penile erection and yawning, the nitric oxide donors sodium nitroprusside, hydroxylamine, isoamyl nitrite and S-nitroso-N-acetyl-penicillamine were injected into the lateral ventricles (i.c.v.) or
M R Melis et al.
European journal of pharmacology, 328(2-3), 115-123 (1997-06-11)
A dose of N-methyl-D-aspartic acid (NMDA, 50 ng) that induces penile erection and yawning when injected into the paraventricular nucleus of the hypothalamus, increased the concentration of NO2- from 1.10 +/- 0.28 microM to 7.32 +/- 1.12 microM and of
T M D'Cunha et al.
Hormones and behavior, 59(1), 14-21 (2010-10-12)
Female rats with maternal experience display a shorter onset of maternal responsiveness compared to those with no prior experience. This phenomenon called 'maternal memory' is critically dependent on the nucleus accumbens (NA) shell. We hypothesized that activation of OT receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service