Skip to Content
Merck
All Photos(2)

Key Documents

O5250

Sigma-Aldrich

D-Ornithine monohydrochloride

~98%

Synonym(s):

(R)-2,5-Diaminopentanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O2 · HCl
CAS Number:
Molecular Weight:
168.62
Beilstein:
4153339
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

D-Ornithine monohydrochloride, ~98%

Assay

~98%

Quality Level

form

powder

color

white

mp

239 °C

SMILES string

Cl[H].NCCC[C@@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m1./s1

InChI key

GGTYBZJRPHEQDG-PGMHMLKASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D-Ornithine cleave proteins at cysteine residues.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan E Cellitti et al.
Nature chemical biology, 7(8), 528-530 (2011-04-29)
D-ornithine has previously been suggested to enhance the expression of pyrrolysine-containing proteins. We unexpectedly discovered that uptake of D-ornithine results in the insertion of a new amino acid, pyrroline-carboxy-lysine (Pcl) instead of the anticipated pyrrolysine (Pyl). Our feeding and biochemical
Sherry Dadsetan et al.
Biochemical pharmacology, 85(1), 115-123 (2012-10-30)
Combined administration of ornithine and phenylacetate (OP) is proposed as a novel treatment of hyperammonemia and hepatic encephalopathy. Ornithine is believed to increase ammonia fixation into glutamine in muscle tissue and glutamine is subsequently thought to react with phenylacetate forming
D Amelio et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 164(2), 356-362 (2012-11-06)
The Frank-Starling law is a fundamental property of the vertebrate myocardium which allows, when the end-diastolic volume increases, that the consequent stretch of the myocardial fibers generates a more forceful contraction. It has been shown that in the eel (Anguilla
Charles D Mills
Critical reviews in immunology, 32(6), 463-488 (2013-02-23)
The purpose of immunology is simple. Cure or prevent disease. M1/M2 is useful because it is simple. M1/M2 describes the two major and opposing activities of macrophages. M1 activity inhibits cell proliferation and causes tissue damage while M2 activity promotes
Irene Tsiapa et al.
Nuclear medicine and biology, 40(2), 262-272 (2012-12-15)
Radiolabeled RGD peptides that specifically target integrin α(ν)β(3) have great potential in early tumor detection through noninvasive monitoring of tumor angiogenesis. Based on previous findings of our group on radiopeptides containing positively charged aminoacids, we developed a new cyclic cRGDfK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service