Skip to Content
Merck
All Photos(2)

Key Documents

N9657

Sigma-Aldrich

Monoclonal Anti-Nitric Oxide Synthase, Inducible antibody produced in mouse

clone NOS-IN, ascites fluid

Synonym(s):

Anti-iNOS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

NOS-IN, monoclonal

mol wt

antigen 130 kDa

contains

15 mM sodium azide

species reactivity

mouse, rat

packaging

antibody small pack of 25 μL

technique(s)

microarray: suitable
western blot: 1:1,000 using whole cell extract of the murine macrophage cell line RAW 264.7 activated with LPS and IFN-γ

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOS2(4843)
mouse ... Nos2(18126)
rat ... Nos2(24599)

General description

Monoclonal Anti-Nitric Oxide Synthase, Inducible (iNOS) (mouse IgG1 isotype) is derived from the NOS-IN hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized BALB/c mouse. Nitric oxide synthase (NOS) is classified into three types based on molecular mass, subcellular location, and Ca2+ dependence. Type I NOS (150-160 kDa) is found in neurons. It is also called as NOS-1, neuronal NOS (nNOS), brain NOS (bNOS), cerebral NOS, constitutive NOS or Ca2+-regulated NOS (cNOS). Type II (130 kDa), best characterized in macrophages, also known as macrophage NOS (mNOS) or inducible NOS (iNOS). Type III (135 kDa) is found in endothelial cells. It is called as endothelial NOS (eNOS, or ecNOS). Type II NOS is Ca2+ -independent and is expressed in activated macrophages and some glial cells after stimulation. iNOS is also found in several other cell types including hepatocytes, chondrocytes, endothelial cells and fibroblasts.

Specificity

Monoclonal Anti-Nitric Oxide Synthase, Inducible (iNOS), antibody reacts specifically with inducible-type nitricoxide synthase (iNOS, also termed macNOS and mNOS). It does not react with NOS derived from brain.

Immunogen

synthetic peptide corresponding to amino acids 1126-1144 of nitric oxide synthase (NOS) from mouse macrophage (iNOS, also termed macNOS) conjugated to KLH. The immunogen sequence is highly conserved in rat iNOS and diverges from the human iNOS.

Application

Monoclonal Anti-Nitric Oxide Synthase, Inducible (iNOS) has been used in
  • immunofluorescence staining
  • immunogold electron microscopy
  • immunoblotting
  • immunohistochemistry
  • immunocytochemistry
  • enzyme linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Nitric oxide synthase (NOS) is an enzyme involved in the synthesis of nitric oxide (NO), a free radical generated under physiological conditions by virtually all mammalian cells. NO is formed from arginine by NOS which oxidizes a guanidino nitrogen of arginine, releasing NO and citrulline. NO is a messenger molecule mediating diverse functions including vasodilatation, neurotransmission, and antimicrobial and anti-tumor activities. Various types of NOS may serve a variety of diverse biological pathways.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jed E Rose et al.
Journal of neuroscience research, 90(4), 849-859 (2012-03-16)
Nicotine in tobacco smoke is thought to stimulate sensory nerve fibers by receptors that are located on airway epithelial cells and on terminal branches of C-fiber afferents, but the exact neurochemical substrate that mediates the sensory effects of nicotine associated
Sapana Bansod et al.
Environmental toxicology, 36(4), 530-539 (2020-11-10)
Borneol is a commonly used flavouring substance in traditional Chinese medicine, which possesses several pharmacological activities including analgesic, antiinflammatory, and antioxidant properties. The aim of this study was to investigate the effects of borneol on cerulein-induced acute pancreatitis (AP) model.
Ultrastructural immunogold localization of nitric oxide synthase isoforms in rat and human eosinophils
Saluja R, et al.
Cell and Tissue Research, 340(2), 381-388 (2010)
Raymond Cespuglio et al.
PloS one, 14(4), e0215070-e0215070 (2019-04-18)
To study the anatomo-biochemical substrates of brain inflammatory processes, Wistar male rats were infected with Trypanosoma brucei brucei. With this reproducible animal model of human African trypanosomiasis, brain cells (astrocytes, microglial cells, neurons) expressing the inducible nitric oxide synthase (iNOS)
Effects of nebulized ketamine on allergen-induced airway hyperresponsiveness and inflammation in actively sensitized Brown-Norway rats
Zhu MM, et al.
Journal of Inflammation, 4(1), 10-10 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service