Skip to Content
Merck
All Photos(1)

Key Documents

N7389

Sigma-Aldrich

3-Nitro-L-tyrosine

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3-4-(OH)CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
226.19
Beilstein:
2813157
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

form

crystalline

Quality Level

mp

233-235 °C (dec.) (lit.)

SMILES string

N[C@@H](Cc1ccc(O)c(c1)[N+]([O-])=O)C(O)=O

InChI

1S/C9H10N2O5/c10-6(9(13)14)3-5-1-2-8(12)7(4-5)11(15)16/h1-2,4,6,12H,3,10H2,(H,13,14)/t6-/m0/s1

InChI key

FBTSQILOGYXGMD-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Nitro-L-tyrosine has been used in:
  • western blot analysis to quantify the nitrotyrosine levels in mice tissue protein
  • Murashige–Skoog medium to treat wild type and mitogen-activated protein kinase 6 (mpk6)-2 mutant seedlings for inferring the connection between nitric oxide (NO) and MPK6 functions in the regulation of mitotic microtubule organization
  • immunocytochemistry

Biochem/physiol Actions

3-Nitro-L-tyrosine is an oxidant and cytotoxic agent. 3-Nitrotyrosine serves as a biological marker of proteins modified by nitrogen-free radical species in systemic autoimmunogenic conditions. It is a marker for oxidative damage mediated by peroxynitrite. Nitrotyrosine is associated with VEGF-C expression and lymph node metastasis in breast cancer. 3-Nitrotyrosine is produced due to oxidative/nitrosative stress-mediated chemical modifications such as nitration of proteins, which is observed in lung carcinogenesis.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eman Ahmed Alaa El-Din et al.
Environmental science and pollution research international, 26(21), 21556-21564 (2019-05-28)
The evaluation of the toxicological effects of titanium dioxide nanoparticles (TiO2NPs) is increasingly important due to their growing occupational and industrial use. Curcumin is a yellow curry spice with a long history of use in herbal medicine and has numerous
Tzu-Hao Li et al.
Scientific reports, 6, 39405-39405 (2016-12-23)
Thalidomide can modulate the TNFα-NFκB and iNOS pathway, which involve in the pathogenesis of hepatopulmonary syndrome (HPS) and muscle wasting in cirrhosis. In bile duct ligated-cirrhotic rats, the increased circulating CD16+ (inflammatory) monocytes and its intracellular TNFα, NFκB, monocyte chemotactic
Jiqu Xu et al.
Journal of pineal research, 67(2), e12584-e12584 (2019-05-03)
Brain insulin resistance, induced by neuroinflammation and oxidative stress, contributes to neurodegeneration, that is, processes that are associated with Aβ accumulation and TAU hyperphosphorylation. Here, we tested the effect of chronic administration of melatonin (MLT) on brain insulin resistance and
Karen S Harris et al.
Scientific reports, 9(1), 10820-10820 (2019-07-28)
Asparaginyl endopeptidases (AEPs) are a class of enzymes commonly associated with proteolysis in the maturation of seed storage proteins. However, a subset of AEPs work preferentially as peptide ligases, coupling release of a leaving group to formation of a new
Remmelt Van der Werf et al.
Cardiovascular diabetology, 17(1), 104-104 (2018-07-22)
Oxidative stress (OS) plays an important role in type 2 diabetes (T2D) pathogenesis and its complications. New therapies target natural antioxidants as an alternative and/or supplemental strategy to prevent and control them. Our previous chemical and biological studies highlighted the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service