Skip to Content
Merck
All Photos(1)

Key Documents

N6288

Sigma-Aldrich

Anti-Nucleolin−Atto 488 antibody produced in rabbit

1.5-3.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NCL, Protein C23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

Atto 488 conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

storage condition

protect from light

concentration

1.5-3.0 mg/mL

technique(s)

direct immunofluorescence: 0.5-1.0 μg/mL using human HeLa, rat NRK and mouse 3T3 cells

fluorescence

λex 500 nm; λem 522 nm in PBS
λex 500 nm; λem 522 nm

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCL(4691)
mouse ... Ncl(17975)
rat ... Ncl(25135)

General description

Nucleolin (NCL) is an anti-apoptotic phosphoprotein. It is highly expressed in the nucleolus as well as other sub-cellular organelles.
Find more information here
Nucleolin is a multi-domain phosphoprotein mapped to human chromosome 2q37. It comprises three domains and a bipartite nuclear localization signal sequence. Nucleolin domains include the highly acidic N-terminal region, a RNA-binding central domain and a glycine, arginine and phenylalanine residues rich C-terminal region.

Specificity

Anti-Nucleolin-Atto 488 recognizes human, rat, and mouse nucleolin.

Immunogen

synthetic peptide corresponding to amino acid residues 2-17 of human nucleolin conjugated to KLH. The corresponding sequence differs by 4 amino acids in rat and mouse nucleolin, and by one amino acid in the rat homolog protein NRP (nucleolin related protein).

Application

Anti-Nucleolin−Atto 488 antibody produced in rabbit has been used in immunocytochemistry and immunofluorescence.

Biochem/physiol Actions

Nucleolin (NCL) interacts with proteins participating in microtubule nucleation. It takes part in the transcription of polymerase I, formation of ribosomes and duplication of centrosomes. NCL is also a transcription regulator. Studies have shown that it is highly expressed in many tumors.
Nucleolin N- and C-terminal domains are involved in protein-protein interaction, whereas the central domain is involved in specific interactions with nucleic acids. Nucleolin exhibits autodegradation, DNA and RNA helicase activities, and DNA-dependent ATPase activity. Nucleolin′s multiple activities may be regulated by proteolysis and methylation. It also interacts with telomerase andthis interaction is critical for the nucleolar localization of telomerase. Nucleolin might function as a cell surface receptor for lipoproteins, viruses, extracellular matrix, growth factors and other molecules.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Fabre et al.
Biomacromolecules, 23(6), 2302-2314 (2022-05-14)
Nucleolin is a multifunctional protein involved in essential biological processes. To precisely localize it and unravel its different roles in cells, fluorescence imaging is a powerful tool, especially super-resolution techniques. Here, we developed polymer-aptamer probes, both small and bright, adapted
Marjan M Tajrishi et al.
Communicative & integrative biology, 4(3), 267-275 (2011-10-08)
Nucleolin is a multifunctional phosphoprotein ubiquitously distributed in the nucleolus, nucleus and cytoplasm of the cell. Nucleolin has a bipartite nuclear localization signal sequence and is conserved in animals, plants and yeast. Its levels are correlated with the rate of
Jian-Yu Xu et al.
Asian Pacific journal of cancer prevention : APJCP, 16(8), 3301-3306 (2015-04-30)
Nucleolin (C23) is an important anti-apoptotic protein that is ubiquitously expressed in exponentially growing eukaryotic cells. In order to understand the impact of C23 in radiation therapy, we attempted to investigate the relationship of C23 expression with the radiosensitivity of
Induced expression of nucleolin phosphorylation-deficient mutant confers dominant-negative effect on cell proliferation.
Xiao S
PLoS ONE, 9(10), e109858-e109858 (2014)
Posterior microphthalmos as a genetically heterogeneous condition that can be allelic to nanophthalmos.
Mohammed A Aldahmesh et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 129(6), 805-807 (2011-06-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service