Skip to Content
Merck
All Photos(1)

Key Documents

MTOXCE2B6

Sigma-Aldrich

CypExpress 2B6 Cytochrome P450 human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

biological source

human

recombinant

expressed in Pichia pastoris

form

dry powder

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP2B6(1555)

Biochem/physiol Actions

CYP2B6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to metabolize some xenobiotics, such as the anti-cancer drugs cyclophosphamide and ifosphamide. Transcript variants for this gene have been described; however, it has not been resolved whether these transcripts are in fact produced by this gene or by a closely related pseudogene, CYP2B7. Both the gene and the pseudogene are located in the middle of a CYP2A pseudogene found in a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q.
Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous.

Features and Benefits

CypExpress 2B6 is a permeabilized and stabilized dried yeast powder preparation containing full length, unmodified, human CYP2B6 and recombinant human NADPH oxidoreductase. Using proprietary processes, CypExpress are expressed in Pichia yeast. Following optimal expression, the cells are inactivated, permeabilized and dried to a fine powder. CypExpress retains the cellular mechanisms to provide the P450 enzymes with the energy and cofactors to continue to function for long experiments, and can generate larger amounts of metabolite than mammalian microsomes or other genetically engineered expression systems.It is designed for rapid screening of drug metabolites and for scaling up metabolite generation.

Legal Information

CypExpress is a trademark of Oxford Biomedical Research

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingkun Song et al.
Scientific reports, 5, 16775-16775 (2015-11-26)
This study investigated interactive effects of CYP2B6 genotypes and liver metastasis on the prognosis of metastatic breast cancer patients who received combined chemotherapy of docetaxel and thiotepa. Totally 153 patients were retrospectively genotyped rs8192719 (c.1294 + 53C > T) and
Jingbao Liu et al.
Biochemistry, 55(13), 1997-2007 (2016-03-17)
Using a combined structural and biochemical approach, the functional importance of a recently described peripheral pocket bounded by the E-, F-, G-, and I-helices in CYP2B4 and 2B6 was probed. Three series of 4-substituted-7-alkoxycoumarin derivatives with -H, -CH3, or -CF3
Esther Ng et al.
Environmental research, 140, 95-101 (2015-04-04)
Polychlorinated biphenyls (PCBs) are a group of man-made environmental pollutants which accumulate in humans with adverse health effects. To date, very little effort has been devoted to the study of the metabolism of PCBs on a genome-wide level. Here, we
Jackson K Mukonzo et al.
Pharmacogenomics, 15(11), 1423-1435 (2014-10-11)
We investigated the effects of rifampicin-based anti-TB treatment on plasma efavirenz exposure and the implications of CYP2B6 genotype. Antiretroviral therapy-naive Ugandan HIV patients without (n = 157) or with TB coinfection (n = 106) were enrolled and treated with efavirenz-based
Séverine Crettol et al.
Clinical pharmacology and therapeutics, 78(6), 593-604 (2005-12-13)
Recent in vitro studies have suggested an important role of cytochrome P450 (CYP) 2B6 and CYP2C19 in methadone metabolism. We aimed to determine the influence of CYP2B6, CYP2C9, and CYP2C19 genetic polymorphism on methadone pharmacokinetics and on the response to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service