Skip to Content
Merck

MTCH6510

MTC Bio Halo pipette tips

capacity 10 mL

Synonym(s):

pipetting tip, MTC Bio H6510, dispensing tip, liquid transfer tip, pipette tips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.02
UNSPSC Code:
41121600
Volume:
10 mL
Sterility:
sterile
Feature:
DNase free, RNase free
Packaging:
pack of 100 ea
Manufacturer/tradename:
MTC Bio H6510
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , polypropylene

sterility

sterile

feature

DNase free, RNase free

packaging

pack of 100 ea

manufacturer/tradename

MTC Bio H6510

capacity

10 mL

volume

10 mL

General description

10mL Pipette Tips for Halo Pipettors, 100/pk
MTC Bio’s Macro 5 and 10mL tips are specially designed for an ideal and comfortable fit on Macro size pipettes. Supplied in bags, these tips are made of ultra-clear polypropylene and are guaranteed to create the most ideal seal on ProPette™ Pipettors.

Features and Benefits

  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips

Legal Information

Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio


Still not finding the right product?

Explore all of our products under MTC Bio Halo pipette tips




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library