Skip to Content
Merck
All Photos(1)

Documents

MSMLS05

Sigma-Aldrich

MSMLS Plate 5 (Water Soluble)

Supplied by IROA Technologies

Synonym(s):

MSMLS, Mass Spectrometry Metabolite Libarary of Standards, Metabolic Library, Metabolite Library

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.28

form

solid

packaging

pkg of 1 plates

manufacturer/tradename

IROA Technologies

plate size

96 wells

shipped in

dry ice

storage temp.

−20°C

General description

MSMLS Plate 5 is a collection of high quality small biochemical molecules that span a broad range of primary metabolism. These are high purity (>95%) compounds supplied in an economical, ready-to-use format. This library of standards is most commonly used for metabolite qualification and quantification for MS and NMR, functional cellular assays, and phenotypic screening. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound, and Substance ID.The library is intended to be used for mass spectrometry metabolomics applications and provides a broad representation of primary metabolism.

Application

Metabolite qualification and quantification for MS, NMR, etc.; Functional cellular assays; Phenotypic screening. Suitable for manual and automated work flows.

Features and Benefits

To view a complete list of compounds in the library for recent lots, download the plate maps below:

The complete plate map with name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMBD ID, PubChem Compound, and Substance ID are provided with the software download.

Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds:

  • Carboxylic acids, amino acids
  • Biogenic amines, polyamines
  • Nucleotides, coenzymes and vitamins
  • Mono- and disaccharides
  • Fatty acids, lipids, steroids and hormones

Preparation Note

MSMLS Plate 5 compounds are conveniently provided at 5 μg per well, enough for multiple injections. Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds by row to allow multiple mixtures to be analyzed per injection. For more information on MSMLS05, solubilization and preparation notes please refer to the product information sheet.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

MLSDiscovery is a trademark of IROA Technologies LLC
MSMLS is a trademark of IROA Technologies LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Repr. 1A - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 - STOT RE 2 - STOT SE 3

Target Organs

Blood, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles M Nichols et al.
Analytical chemistry, 90(24), 14484-14492 (2018-11-20)
In this work, we established a collision cross section (CCS) library of primary metabolites based on analytical standards in the Mass Spectrometry Metabolite Library of Standards (MSMLS) using a commercially available ion mobility-mass spectrometer (IM-MS). From the 554 unique compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service