Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.75
UNSPSC Code:
12352207
Quality Segment
recombinant
expressed in E. coli Taq
form
liquid, lyophilized
parameter
2 μL sample volume
shipped in
ambient
storage temp.
2-8°C
General description
Lookout® One-Step Mycoplasma PCR Detection Kit is a kit for direct detection of mycoplasma contaminations in cell cultures, cell culture media, and other biological matrices by conventional PCR. Designed to require minimal pipetting, the kit includes all reagents required for PCR: primers, nucleotides, polymerase and the internal amplification control, which are provided in a ready-to-use lyophilized reaction mix. Upon rehydration with the included buffer, the mix is ready to be dispensed into the reaction tubes according to the sample number. Only samples or controls need to be added prior to PCR.
Application
Applicable in research and industry for direct testing of cell cultures and biologicals. Intended for research use only. Not recommended for clinical diagnostics or testing of human samples.
Biochem/physiol Actions
Type of PCR:
For conventional, endpoint PCR with gel electrophoresis
Result Evaluation: Gel electrophoresis at endpoint of PCR
For conventional, endpoint PCR with gel electrophoresis
Result Evaluation: Gel electrophoresis at endpoint of PCR
Preparation Note
Lyophilized OneStep Mix is reconstituted in 600μl Rehydration Buffer; Lyophilized Positive Control DNA is reconstituted in 300μl PCR Grade Water
Store the unopened components at +2 °C to +8 °C until the expiry date indicated on the label. Once rehydrated, the components must be stored at ≤ -18 °C.
Other Notes
Kit Components:
Lyophilized Mix (Primer sets / Nucleotides / Internal Control DNA and polymerase at optimized concentrations in aliquots of 25 or 100 reactions each Rehydration Buffer)
Lyophilized Positive Control DNA
PCR Grade Water
Lyophilized Mix (Primer sets / Nucleotides / Internal Control DNA and polymerase at optimized concentrations in aliquots of 25 or 100 reactions each Rehydration Buffer)
Lyophilized Positive Control DNA
PCR Grade Water
Required Consumables:
Agarose gels and DNA stains
PCR reaction tubes and pipette filter tips
Required Lab Devices:
PCR cycler for conventional PCR
Gel electrophoresis and documentation system
Pipetting equipment
Microcentrifuge
Agarose gels and DNA stains
PCR reaction tubes and pipette filter tips
Required Lab Devices:
PCR cycler for conventional PCR
Gel electrophoresis and documentation system
Pipetting equipment
Microcentrifuge
Legal Information
LookOut is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.