Skip to Content
Merck
All Photos(1)

Key Documents

MAK224

Sigma-Aldrich

Fatty Acid Methylation Kit

Manufactured by Nacalai Tesque, Inc.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 100 tests

application(s)

cosmetics
food and beverages
sample preservation

storage temp.

room temp

General description

Methyl esterification of fatty acids is commonly done prior to gas chromatography analysis to prevent peak tailing and to increase sample volatility. However, the conventional esterification procedure requires specialized equipment and high technical skill. By using the Fatty Acid Methylation kit that employs a new reaction technique, followed by the fatty acid methyl ester purification kit (MAK225), fatty acid methyl esterification is greatly simplified.

Suitability

Suitable for the methyl esterification of fatty acids (glycerolipids such as triglycerides, phospholipids, glycolipids, lecithin and sterol esters). Samples include blood, yeast, liver, cooking oil, soybean powder, fish oil, etc.

Principle

The Fatty Acid Methylation Kit followed by the Fatty Acid Methyl Ester Purification Kit (Catalog Number MAK225) employs a proprietary reaction technique. Using these kits, the fatty acid methyl esterification/purification process is greatly simplified.

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Repr. 2 - Self-heat. 1 - Skin Corr. 1A - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes, Nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

Flash Point(F)

-14.8 °F

Flash Point(C)

-26 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriella Leone et al.
Scientific reports, 11(1), 5209-5209 (2021-03-06)
Diatom microalgae have great industrial potential as next-generation sources of biomaterials and biofuels. Effective scale-up of their production can be pursued by enhancing the efficiency of their photosynthetic process in a way that increases the solar-to-biomass conversion yield. A proof-of-concept

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service