Skip to Content
Merck
All Photos(1)

Key Documents

MAK113

Sigma-Aldrich

ATPase/GTPase Activity Assay Kit

sufficient for 200 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 200 colorimetric tests

detection method

colorimetric

relevant disease(s)

cancer

storage temp.

2-8°C

General description

ATPases and GTPases catalyze the decomposition of ATP or GTP into ADP or GDP and free phosphate. These enzymes play key roles in transport, signal transduction, protein biosynthesis, and cell differentiation.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of ATPase and GTPase activity and for the screening of ATPase and GTPase inhibitors.

Principle

The ATPase/GTPase Activity Assay kit provides a simple and direct procedure for measuring ATPase/GTPase activity in a microplate format. This kit uses a single reagent formulation to accurately determine enzyme activity in 30 minutes at room temperature. The malachite green reagent forms a stable dark green color with free phosphate liberated by the enzymes resulting in a colorimetric product, measured at 620 nm (600-660 nm), proportional to the enzyme activity present One unit of activity is the amount of enzyme that catalyzes the production of 1 μmole of free phosphate per minute under the assay conditions.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng-Yan Sun et al.
Oncology reports, 39(6), 2673-2680 (2018-04-06)
Regulator of G‑protein signaling 1 (RGS1) has been found to be a critical factor in melanoma and other malignancies. However, the mechanism involved in the RGS1‑mediated promotion of melanoma progression is not clear. We based our study on samples collected from
Shannon E Hill et al.
PloS one, 12(7), e0180241-e0180241 (2017-07-26)
Dihydroneopterin triphosphate pyrophosphatase (DHNTPase), a member of the Mg2+ dependent Nudix hydrolase superfamily, is the recently-discovered enzyme that functions in the second step of the pterin branch of the folate biosynthetic pathway in E. coli. DHNTPase is of interest because
Xindan Wang et al.
Molecular cell, 71(5), 841-847 (2018-08-14)
Structural maintenance of chromosomes (SMC) complexes shape the genomes of virtually all organisms, but how they function remains incompletely understood. Recent studies in bacteria and eukaryotes have led to a unifying model in which these ring-shaped ATPases act along contiguous
Emilie Mahieu et al.
Biophysical journal, 119(2), 375-388 (2020-07-09)
The proteasome is a key player of regulated protein degradation in all kingdoms of life. Although recent atomic structures have provided snapshots on a number of conformations, data on substrate states and populations during the active degradation process in solution
Shuhui Wang et al.
Protein & cell, 11(2), 124-137 (2019-11-24)
Type VII secretion systems (T7SSs) are found in many disease related bacteria including Mycobacterium tuberculosis (Mtb). ESX-1 [early secreted antigen 6 kilodaltons (ESAT-6) system 1] is one of the five subtypes (ESX-1~5) of T7SSs in Mtb, where it delivers virulence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service