M7316
Monoamine Oxidase A human
recombinant, expressed in baculovirus infected BTI insect cells
Synonym(s):
MAO-A, MAOA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in baculovirus infected BTI insect cells
Quality Level
form
liquid
packaging
vial of ~2.5 mg
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... MAOA(4128)
General description
Monoamine Oxidase A contains binding sites for 8α-S-cysteinyl-FAD.
Monoamine oxidases metabolize biogenic amines in the central nervous system and the peripheral tissue.
Application
Monoamine Oxidase A has been used in a study to assess abnormal behavior in a large kindred of males where a deficiency of enzymatic activity of monamine oxidase A was found. It has also been used in a study to investigate an association between smoking and the inhibition of MAOA.
Biochem/physiol Actions
MAO′s are proteins of the mitochondrial membrane. These enzymes are responsible for catalyzing oxidative deamination of endo- and xenobiotic amines. Substrate specificity differs for each isozyme.
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Proceedings of the National Academy of Sciences of the United States of America, 93(24), 14065-14069 (1996-11-26)
Several studies have documented a strong association between smoking and depression. Because cigarette smoke has been reported to inhibit monoamine oxidase (MAO) A in vitro and in animals and because MAO A inhibitors are effective antidepressants, we tested the hypothesis
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 62(2), 253-258 (2020-07-11)
Reactive astrocytes play a key role in the pathogenesis of various neurodegenerative diseases. Monoamine oxidase-B (MAO-B) is one of the promising targets for the imaging of astrogliosis in the human brain. A novel selective and reversible MAO-B tracer, (S)-(2-methylpyrid-5-yl)-6-[(3-18F-fluoro-2-hydroxy)propoxy]quinoline (18F-SMBT-1)
Antioxidants (Basel, Switzerland), 10(10) (2021-10-24)
Neurotransmitter catecholamines (dopamine, epinephrine, and norepinephrine) are liable to undergo oxidation, which copper is deeply involved in. Catecholamine oxidation-derived neurotoxicity is recognized as a pivotal pathological mechanism in neurodegenerative diseases. Glutamate, as an excitatory neurotransmitter, is enriched in the brain
Analytical and bioanalytical chemistry, 408(3), 695-703 (2015-11-18)
The need for rapid and cost-effective pre-screening protocols of the toxicological response of the vast array of emerging nanoparticle types is apparent and the emerging consensus on the paradigm of oxidative stress by generation of intracellular reactive oxygen species as
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(3), 460-465 (2014-02-15)
Serotonergic biosynthesis in the endocrine pancreas, of which the islets of Langerhans is the major constituent, has been implicated in insulin release and β cell proliferation. In this study, we investigated the feasibility of quantitative noninvasive imaging of the serotonergic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service