Skip to Content
Merck
All Photos(2)

Key Documents

M7033

Sigma-Aldrich

NG-Methyl-L-arginine acetate salt

≥98% (TLC), powder, nitric oxide synthase inhibitor

Synonym(s):

Nω-Methyl-L-arginine acetate salt, L-NMMA, N5-(Methylamidino)-L-ornithine acetate salt, NG-Monomethyl-L-arginine acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N4O2 · C2H4O2
CAS Number:
Molecular Weight:
248.28
Beilstein:
6674255
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

Product Name

NG-Methyl-L-arginine acetate salt, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

water: 49.00-51.00 mg/mL, clear to very slightly hazy, colorless to faintly yellow

SMILES string

CC(O)=O.CNC(=N)NCCC[C@H](N)C(O)=O

InChI

1S/C7H16N4O2.C2H4O2/c1-10-7(9)11-4-2-3-5(8)6(12)13;1-2(3)4/h5H,2-4,8H2,1H3,(H,12,13)(H3,9,10,11);1H3,(H,3,4)/t5-;/m0./s1

InChI key

IKPNWIGTWUZCKM-JEDNCBNOSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

NG-Methyl-L-arginine acetate salt has been used:
  • as an inhibitor of nitric oxide synthase (NOS) in wound healing studies in injured mice
  • as an inhibitor of nitric oxide synthase (NOS) to study nitric oxide (NO) immunosuppressive effect of murine stem cells
  • as an inhibitor to monitor NO levels in hemocytes respiratory burst assay

Biochem/physiol Actions

NG-Methyl-L-arginine is an inhibitor of nitric oxide synthase in neurons, endothelial cells, and macrophages. It inhibits endothelium-derived relaxing factor and promotes contraction in pulmonary artery and vein. It stereo selectively inhibits the generation of nitric oxide (NO) from arginine. It promotes epinephrine and glucose levels, contributing to hyperglycemia in central nervous system.

Features and Benefits

This compound is featured on the Nitric Oxide Synthases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safa Al Therwani et al.
BMC nephrology, 18(1), 86-86 (2017-03-16)
Tolvaptan is a selective vasopressin receptor antagonist. Nitric Oxide (NO) promotes renal water and sodium excretion, but the effect is unknown in the nephron's principal cells. In a dose-response study, we measured the effect of tolvaptan on renal handling of
NG-methyl-L-arginine, an inhibitor of nitric oxide synthase, affects the central nervous system to produce peripheral hyperglycemia in conscious rats
Uemura K, et al.
Neuroendocrinology, 66(2), 136-144 (1997)
Immune parameters in the common octopus (Octopus vulgaris Cuvier, 1797) naturally infected by the gastrointestinal protozoan parasite Aggregata octopiana
Castellanos-Martinez S and Gestal C
Aquaculture (Amsterdam, Netherlands), 497, 487-493 (2018)
NG-methyl-L-arginine functions as an alternate substrate and mechanism-based inhibitor of nitric oxide synthase
Olken NM and Marletta MA
Biochemistry, 32(37), 9677-9685 (1993)
Nitric oxide induces epidermal stem cell de-adhesion by targeting integrin beta1 and Talin via the cGMP signalling pathway
Zhan R, et al.
Nitric Oxide, 78, 1-10 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service