Skip to Content
Merck
All Photos(2)

Key Documents

M6818

Sigma-Aldrich

Anti-MeCP2 antibody, Mouse monoclonal

clone Mec-168, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

Mec-168, monoclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.1-0.2 μg/mL using nuclear cell extract of cultured Jurkat, human acute T cell leukemia cells, or cultured MCF7 human breast carcinoma.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MECP2(4204)
mouse ... Mecp2(17257)
rat ... Mecp2(29386)

General description

Monoclonal Anti-MeCP2 (mouse IgG1 isotype) is derived from the Mec-168 hybridoma produced by the fusion of mouse myeloma cells (NS1) and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to the C- terminus of human MeCP2. Methyl-CpG binding protein 2 (MeCP2) is the first methyl-CpG-binding protein to be isolated. This protein contains a methyl-CpG-binding domain (MBD) and a transcriptional repression domain (TRD).

Immunogen

synthetic peptide corresponding to the C-terminus (amino acids 471-486) of human MeCP2.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)
Western Blotting (1 paper)
Monoclonal Anti-MeCP2 antibody produced in mouse has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunostaining
  • enzyme linked immunosorbent assay (ELISA)
  • immunocytochemistry
  • western blot (0.1-0.2 μg/mL using nuclear cell extract of cultured Jurkat, human acute T cell leukemia cells, or cultured MCF7 human breast carcinoma)

Biochem/physiol Actions

Methyl-CpG binding protein 2 (MeCP2) is a transcription modulator that binds methylated DNA. This protein regulates neuronal functions and central nervous system development. Alterations in MeCP2 have been associated with neurological diseases such as MECP2 duplication syndrome and Rett syndrome
Methyl-CpG binding protein 2 (MeCP2) deacetylase (HDAC) repressive machinery via recruitment of the Sin 3A corepressor, thus removing acetyl groups from histones and consequently, silencing genes. Antibodies reacting specifically with MeCP2 may be used for studying chromatin remodeling effects on gene expression.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rett syndrome mutations abolish the interaction of MeCP2 with the NCoR/SMRT co-repressor
Lyst MJ, et al.
Nature Neuroscience, 16(7) (2013)
The biological functions of the methyl-CpG-binding protein MeCP2 and its implication in Rett syndrome
Nan X, et al.
Brain & Development, 23, S32-S37 (2001)
Jacky Guy et al.
Annual review of cell and developmental biology, 27, 631-652 (2011-07-05)
Methyl-CpG binding protein 2 (MeCP2) was first identified in 1992 as a protein that binds specifically to methylated DNA. Mutations in the MECP2 gene were later found to be the cause of an autism spectrum disorder, Rett syndrome. Despite almost
Oz Pomp et al.
Cell stem cell, 9(2), 156-165 (2011-08-06)
Somatic tissues in female eutherian mammals are mosaic due to random X inactivation. In contrast to mice, X chromosome reactivation does not occur during the reprogramming of human female somatic cells to induced pluripotent stem cells (iPSCs), although this view
The molecular basis of variable phenotypic severity among common missense mutations causing Rett syndrome
Brown K, et al.
Human Molecular Genetics, 25(3), 558-570 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service