All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H6N4
CAS Number:
Molecular Weight:
134.14
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Recommended Products
biological source
synthetic (organic)
Assay
≥99%
form
powder
mp
237-238 °C (lit.)
solubility
water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow
storage temp.
−20°C
SMILES string
Cc1ncnc2[nH]cnc12
InChI
1S/C6H6N4/c1-4-5-6(9-2-7-4)10-3-8-5/h2-3H,1H3,(H,7,8,9,10)
InChI key
SYMHUEFSSMBHJA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
6-Methylpurine (MeP) is a toxic adenine analog used as a bisubstrate inhibitor of enzymes that bind adenosyl moieties and which may become phosphorylated to levels that inhibit RNA and protein synthesis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Paromita Chakraborty et al.
Environmental pollution (Barking, Essex : 1987), 249, 1071-1080 (2019-05-31)
The Hooghly River (HR) estuary is the first deltaic off-shoot of the perennial and transboundary river, Ganga, India. HR receives industrial and domestic waste along with storm-water run-off from Kolkata city and the adjoining districts. Organic micropollutants (OMPs) have been
L J Lockett et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 3(11), 2075-2080 (1998-11-17)
Enzyme-prodrug therapy for the treatment of cancer is an experimental procedure that is under intensive investigation. However, the relative merits of the various systems for use under specific conditions are still being determined. We have compared the efficacy of cell
Hoonsik Yoom et al.
The Science of the total environment, 634, 677-686 (2018-04-12)
The reaction kinetics, products, and pathways of methylparaben (MeP) during water chlorination with and without bromide (Br-) were investigated to better understand the fate of parabens in chlorinated waters. During the chlorination of MeP-spiked waters without Br-, MeP was transformed
Yingxin Yu et al.
Ecotoxicology and environmental safety, 182, 109419-109419 (2019-07-14)
Parabens are a kind of preservatives widely used in cosmetic and personal care products and ubiquitously detected in the environment. However, little is known on human exposure to these chemicals. Our study mainly investigated the urinary parabens in adults from
H Ohgaki et al.
Journal of cancer research and clinical oncology, 117(1), 13-18 (1991-01-01)
The formation of O6-methyldeoxyguanosine (O6-MedGuo) was determined by an immuno-slot-blot assay in DNA of various tissues of F344 rats exposed to N-methyl-N-nitrosourea (MNU) in the drinking water at 400 ppm for 2 weeks. Although the pyloric region of the glandular
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service