Skip to Content
Merck
All Photos(1)

Key Documents

M6403

Sigma-Aldrich

(±)-3,4-Methylenedioxymethamphetamine hydrochloride

Synonym(s):

(±)-MDMA hydrochloride, DL-3,4-Methylenedioxymethamphetamine, DL-MDMA hydrochloride, Ecstasy, XTC hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2 · HCl
CAS Number:
Molecular Weight:
229.70
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: soluble

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

SMILES string

Cl[H].CNC(C)Cc1ccc2OCOc2c1

InChI

1S/C11H15NO2.ClH/c1-8(12-2)5-9-3-4-10-11(6-9)14-7-13-10;/h3-4,6,8,12H,5,7H2,1-2H3;1H

InChI key

LUWHVONVCYWRMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hallucinogen; releases serotonin and dopamine from axon terminals; selective serotonergic neurotoxin.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahra Nazari et al.
Iranian journal of basic medical sciences, 25(9), 1091-1096 (2022-10-18)
Ecstasy is a popular recreational psychostimulant with side effects on the central nervous system. This study examined the corpus striatum tissue of adult rats that received ecstasy during the embryonic period for histological and molecular studies. Rats were divided into
M M Iravani et al.
Synapse (New York, N.Y.), 36(4), 275-285 (2000-05-20)
We examined the effects of pressure ejected 3, 4-methylenedioxymethamphetamine (MDMA) from a micropipette on direct chemically stimulated release, and on electrically stimulated serotonin (5-HT) or dopamine (DA) release in the caudate putamen (CPu), nucleus accumbens (NAc), substantia nigra pars reticulata
G A Ricaurte et al.
Neuropsychobiology, 42(1), 5-10 (2000-06-27)
The popular recreational drug, (+/-)3, 4-methylenedioxymethamphetamine (MDMA; 'Ecstasy') is a potent and selective brain serotonin (5-HT) neurotoxin in animals. MDMA-induced 5-HT neurotoxicity can be demonstrated using a variety of neurochemical, neuroanatomical and, more recently, functional measures of 5-HT neurons. Although
A Lavelle et al.
British journal of pharmacology, 128(5), 975-980 (1999-11-11)
1. We have investigated the effects of methylenedioxymethamphetamine (MDMA, 'ecstasy') on peripheral noradrenergic neurotransmission in the rat. 2. In rat atrial slices pre-incubated with [3H]-noradrenaline and in the presence of desipramine (1 micronM) to prevent effects of MDMA on basal
M I Colado et al.
British journal of pharmacology, 126(4), 911-924 (1999-04-08)
1. We investigated whether dopamine plays a role in the neurodegeneration of 5-hydroxytryptamine (5-HT) nerve endings occurring in Dark Agouti rat brain after 3,4-methylenedioxymethamphetamine (MDMA or 'ecstasy') administration. 2. Haloperidol (2 mg kg(-1) i.p.) injected 5 min prior and 55

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service