Skip to Content
Merck
All Photos(1)

Documents

M5353

Sigma-Aldrich

Mitomycin C

liquid, non-animal origin, suitable for cell culture, BioReagent

Synonym(s):

Ametycine, MMC, MitoExtra, Mitocin C, Mitomycin, Mitonco, Mitoplus, Mitozytrex, Mutamycin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.76

product name

Mitomycin C ready made solution, 10 mg/mL in DMSO, ≥98% (HPLC), Cell culture tested

Quality Level

Assay

≥98% (HPLC)

form

liquid

storage condition

protect from light

concentration

10 mg/mL in DMSO

storage temp.

−20°C

General description

Mitomycin C (MMC) produced by Streptomyces caespitosus is an anticancer antibiotic agent which inhibits proliferation of various cell types in vitro. MMC is DNA alkylating agent, that causes cross linking of complementary DNA strands resulting in: mutagenesis, inhibition of DNA synthesis, initiation of DNA repair events, and activation of apoptosis. Furthermore, MMC has an effect on ribosomal RNA leading to protein synthesis inhibition.
MMC is capable of inhibiting cell proliferation of human or murine embryonic fibroblast cells at concentration range of 0.2-20μg/mL. These treated cells are subsequently used as an adherent feeder layer in long-term culture assays mainly for growth of pluripotent stem cells. Feeder layer cells support the target cells by releasing growth factors to the cell culture and detoxification of the medium. Moreover, feeder layer cells play a role in ECM proteins synthesis that controls cell culture growth. In addition, feeder layer cells are used as a substrate for attachment of the target cells.
MMC also was used for BLC (basal-like cancer) cell line.
MMC as antibiotic agent showed a potent activity against MDR gram-negative bacteria like P. aeruginosa (2 μg/mL), E.Coli (0.5μg/mL) and A. baumannii (16μg/mL). MMC also has a strong activity against gram positive bacteria (0.2μg/mL).

Technical information
1.Mitomycin C ready made solution provides a safer alternative to the powder form by preventing hazardous particles exposure.
2.Please avoid repeated freeze thaw cycles of this product.
3.Mitomycin C solutions are light sensitive and should kept in the dark.
4.It is recommended to keep Mitomycin C solutions at pH 6-7.
5.The recommended concentration of Mitomycin C in cell culture assays is 0.2-20μg/mL. (Ref.8,9,12,13) Therefore, it is recommended to dilute the ready made solution 1:500-50,000.

Caution

Light sensitive

Physical form

Liquid - In DMSO

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ponchio et al.
Cytotherapy, 2(4), 281-286 (2002-06-04)
Mitomycin C (MMC), an antitumoral antibiotic, has been described inhibiting the proliferation of different cell types in vitro. Since irradiation is commonly used to stop the cell growth of adherent cells in several experimental models, we aimed to define the
Ronald Domalaon et al.
Frontiers in microbiology, 10, 1556-1556 (2019-07-30)
The lack of therapeutic options to treat infections caused by multidrug-resistant (MDR) pathogens, especially Gram-negative bacteria, is apparent. Therefore, it is imperative to develop new strategies to address the problem of antimicrobial resistance. Repurposing non-antibiotic commercial drugs for antimicrobial therapy
Mitomycin, a new antibiotic from Streptomyces. I.
T HATA et al.
The Journal of antibiotics, 9(4), 141-146 (1956-07-01)
Ryan G Snodgrass et al.
The Journal of biological chemistry, 285(25), 19068-19075 (2010-04-27)
Mitomycin C (MMC) is a commonly used and extensively studied chemotherapeutic agent requiring biological reduction for activity. Damage to nuclear DNA is thought to be its primary mechanism of cell death. Due to a lack of evidence for significant MMC
M Namba et al.
In vitro, 18(5), 469-475 (1982-05-01)
The effect of feeder layers on cloning efficiency of transformed human cells was investigated. Embryonic human skin or lung fibroblasts; adult human skin fibroblasts; early passage cells from embryos of mouse, rat, and hamster; established mouse cell lines; 3T3 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service