M5170
IgM, Lambda from murine myeloma
clone MOPC 104E, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Mouse IgM-λ
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
clone
MOPC 104E, monoclonal
form
buffered aqueous solution
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
IgM antibodies are present as pentamers in the serum and are produced in response to antigens
IgM, λ from murine myeloma is produced by MOPC 104E tumor line introduced subcutaneously in BALB/c mice. It specifically recognizes mouse IgM, λ. Identity and purity of the immunoglobulin is established by immunoelectrophoresis.
IgM, λ from murine myeloma is produced by MOPC 104E tumor line introduced subcutaneously in BALB/c mice. It specifically recognizes mouse IgM, λ. Identity and purity of the immunoglobulin is established by immunoelectrophoresis.
Specificity
Antigen Specificity: α(1→3) glucose
Application
IgM, λ from murine myeloma was used as standard in ELISA and as isotype control in immunohistochemistry applications.
Biochem/physiol Actions
IgM antibodies are produced initially during B-cell development. IgM participates in opsonization and compliment fixation. Defect in IgM production is observed in X-linked agammaglobulinemia.
Physical form
Solution in 0.05 M Tris, 0.5 M sodium chloride, pH 8.0, containing 0.02% sodium azide
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
D R Wilson et al.
Protein engineering, 10(5), 531-540 (1997-05-01)
The immunodominant region of the Plasmodium falciparum circumsporozoite protein is comprised mainly of a series of tetrapeptide repeats that can, depending on the starting cadence chosen, be described as (NANP)n, (ANPN)n, (NPNA)n or (PNAN)n in one-letter amino acid code. Data
Jason R Nesbitt et al.
Frontiers in microbiology, 8, 2706-2706 (2018-02-07)
Aspergillus fumigatus is a filamentous fungus that can cause a life-threatening invasive pulmonary aspergillosis (IPA) in immunocompromised individuals. We previously characterized an exo-sialidase from A. fumigatus that prefers the sialic acid substrate, 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (Kdn); hence it is a Kdnase.
Establishment of fetal bovine intestinal epithelial cell cultures susceptible to bovine rotavirus infection
Kaushik RS et al
J. Virol. Methods, 148, 182-196 (2008)
Pratik Katwal et al.
Cytotechnology, 71(1), 127-148 (2019-01-03)
The intestinal epithelium is a major site of interaction with pathogens. In bovine intestinal epithelial cells (BIECs), Toll-like receptors (TLRs) play an important role in innate immune responses against enteric pathogens. This study is aimed at establishing a stable bovine
Chithra C Sreenivasan et al.
Virology, 528, 152-163 (2019-01-08)
Influenza viruses are a group of respiratory pathogens that have evolved into four different types: A, B, C, and D. A common feature is that all four types are capable of replicating and transmitting among pigs. Here, we describe the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service