Skip to Content
Merck
All Photos(2)

Key Documents

M4528

Sigma-Aldrich

Monoclonal Anti-MAP1b antibody produced in mouse

clone AA6, ascites fluid

Synonym(s):

Anti-MAP1.2, Anti-MAP1X, Anti-MAP5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ARR
WB
clone:
AA6, monoclonal
species reactivity:
human, feline, bovine, hamster, chicken, mouse, rat
citations:
11
technique(s):
microarray: suitable
western blot: 1:500 using a fresh total rat brain extract or an enriched microtubule protein preparation

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AA6, monoclonal

contains

15 mM sodium azide

species reactivity

human, feline, bovine, hamster, chicken, mouse, rat

technique(s)

microarray: suitable
western blot: 1:500 using a fresh total rat brain extract or an enriched microtubule protein preparation

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP1B(4131)
mouse ... Mtap1b(17755)
rat ... Map1b(29456)

General description

MAP1b is a microtubule associated protein that interacts with actin filaments. MAP1b regulates microtubule stabilization and the development of axons. It may also act as a scaffolding protein Addition of antibody to microtubule proteins before polymerization in immunoassays results in abnormally short (but otherwise morphologically normal) microtubules. Immunohistochemical staining of brain tissue with the product shows selective labeling of dendritic trees throughout the brain. Monoclonal Anti-MAP1b does not react with tubulin of other microtubule associated proteins.
Monoclonal Anti-MAP1b (MAP5) (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Microtubule-associated protein (MAP1B) is the major microtubule associated protein in developing brain which changes its expression during development. In the new born rat brain, it is a major component of microtubules but in the adult its level is ten-fold lower.

Specificity

Monoclonal Anti-MAP1b antibody is specific for MAP1b in humans, mice, rats, bovines, cats, hamsters and chickens. The antibody does not cross-react with other MAPs or tubulin. Addition of the antibody to microtubule proteins before polymerization results in abnormally short (but otherwise morphologically normal) microtubules. In immunohistochemical staining of brain tissue, the antibody shows selective labeling of neurons with stronger staining of axons, dendrites and cell bodies.

Immunogen

rat brain microtubule-associated proteins (MAPs)

Application

Monoclonal Anti-MAP1b antibody produced in mouse has been used in:
  • immunohistochemistry
  • immunostaining
  • fluorescence microscopy

Biochem/physiol Actions

Microtubule-associated protein (MAP1B) is the first MAP to appear in growing axons during development as it is present from the first emergence of the nascent axon from the cell body. Monoclonal Anti-MAP1b may be used to study MAP expression and cytological localization both in tissues and cell lines, under different developmental and environmental circumstances.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of microtubule-associated protein (MAP1B) with growing axons in cultured hippocampal neurons
Fischer I and Romano CG
Molecular and Cellular Neurosciences, 2(1), 39-51 (1991)
Human mesenchymal precursor cells (Stro-1+) from spinal cord injury patients improve functional recovery and tissue sparing in an acute spinal cord injury rat model
Hodgetts SI, et al.
Cell Transplantation, 22(3), 393-412 (2013)
Masamitsu Shimazawa et al.
The Journal of pharmacology and experimental therapeutics, 332(2), 380-387 (2009-11-17)
We examined whether ((1S)-1-((((1S)-1-benzyl-3-cyclopropylamino-2,3-di-oxopropyl)amino)carbonyl)-3-methylbutyl)carbamic acid 5-methoxy-3-oxapentyl ester (SNJ-1945), a new orally available calpain inhibitor, might reduce retinal cell death in vivo and/or in vitro. Retinal cell damage was induced in vivo in mice by intravitreal injection of N-methyl-d-aspartate (NMDA), and
Carolina Paola García et al.
PloS one, 11(3), e0152607-e0152607 (2016-04-01)
Human embryonic stem cells (hESCs) are hypersensitive to genotoxic stress and display lower survival ability relative to their differentiated progeny. Herein, we attempted to investigate the source of this difference by comparing the DNA damage responses triggered by the topoisomerase
Carolina Montenegro-Venegas et al.
Molecular biology of the cell, 21(20), 3518-3528 (2010-08-20)
Cultured neurons obtained from MAP1B-deficient mice have a delay in axon outgrowth and a reduced rate of axonal elongation compared with neurons from wild-type mice. Here we show that MAP1B deficiency results in a significant decrease in Rac1 and cdc42

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service