Skip to Content
Merck
All Photos(1)

Documents

M4145

Sigma-Aldrich

Minoxidil

≥99% (TLC), powder, androgenetic alopecia drug

Synonym(s):

6-(1-Piperidinyl)-2,4-pyrimidinediamine 3-oxide, 6-(1-Piperidinyl)pyrimidine-2,4-diamine 3-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N5O
CAS Number:
Molecular Weight:
209.25
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Minoxidil, ≥99% (TLC)

Assay

≥99% (TLC)

mp

272-274 °C (dec.) (lit.)

originator

Johnson & Johnson

SMILES string

NC1=CC(=NC(=N)N1O)N2CCCCC2

InChI

1S/C9H15N5O/c10-7-6-8(12-9(11)14(7)15)13-4-2-1-3-5-13/h6,11,15H,1-5,10H2

InChI key

ZIMGGGWCDYVHOY-UHFFFAOYSA-N

Gene Information

human ... KCNJ1(3758)

Looking for similar products? Visit Product Comparison Guide

Application

Minoxidil has been used as test drug:
  • in dermal papilla spheroid model, to study the effect of hair regeneration
  • administered orally to experimental rats, in the approach to identify biomarkers of drug induced vascular injury
  • administered via oral gavage to nonhuman primate model in order to study its effect on tilt responses

Biochem/physiol Actions

Minoxidil is originally used to treat hypertension, which also induces hypertrichosis. It is considered as a potent drug for treating androgenetic alopecia.
Activates ATP-activated K+ channels; vasodilator; slow or stop hair loss and promote hair regrowth.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J I Kourie
The Journal of membrane biology, 164(1), 47-58 (1998-06-24)
The lipid bilayer technique was used to examine the effects of the ATP-sensitive K+ channel inhibitor (glibenclamide) and openers (diazoxide, minoxidil and cromakalim) and Cl- channel activators (GABA and diazepam) on two types of chloride channels in the sarcoplasmic reticulum
Ilaria Aurigemma et al.
Biochemical and biophysical research communications, 720, 150104-150104 (2024-05-16)
The T-BOX transcription factor TBX1 is essential for the development of the pharyngeal apparatus and it is haploinsufficient in DiGeorge syndrome (DGS), a developmental anomaly associated with congenital heart disease and other abnormalities. The murine model recapitulates the heart phenotype
Stefan Sarkovich et al.
Journal of experimental orthopaedics, 10(1), 84-84 (2023-08-22)
Synovial fibrosis (SFb) formation and turnover attributable to knee osteoarthritis (KOA) can impart painful stiffness and persist following arthroplasty. To supplement joint conditioning aimed at maximizing peri-operative function, we evaluated the antifibrotic effect of Minoxidil (MXD) on formation of pyridinoline
Establishment of an in vitro organoid model of dermal papilla of human hair follicle
Gupta AC, et al.
Journal of Cellular Physiology, 233(11), 9015-9030 (2018)
J van der Velden et al.
Cellular and molecular life sciences : CMLS, 55(5), 788-798 (1999-06-24)
To investigate whether during cardiac hypertrophy changes occur in contractile protein composition and in mechanical and energetic properties of the myocardium, contractile protein composition, isometric force and adenosine triphosphate (ATP) consumption were studied in control and hypertrophied guinea-pig hearts. Cardiac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service