Skip to Content
Merck
All Photos(2)

Key Documents

M2800

Sigma-Aldrich

A2F Glycan ammonium salt

from Porcine thyroglobulin, ≥90%

Synonym(s):

(NeuNAc-Gal-GlcNAc)2Man3(Fuc)(GlcNAc)2, Disialylated, galactosylated, fucosylated biantennary N-linked glycan, Mannotriose-(fucosyl-di-[N-acetylglucosamine]), bi(sialyl-galactosyl-N-acetylglucosaminyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C90H148N6O66
CAS Number:
Molecular Weight:
2370.14
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Porcine thyroglobulin

Quality Level

Assay

≥90%

storage temp.

−20°C

General description

A2F Glycan is a disialylated-galactosylated biantennary, core-substituted with fucose glycan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service