Skip to Content
Merck
All Photos(1)

Key Documents

M0285

Sigma-Aldrich

Methyl-β-D-galactopyranoside

Synonym(s):

Methyl β-D-galactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
Beilstein:
81569
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥98% (GC)

form

powder

technique(s)

gas chromatography (GC): suitable

color

white to faint yellow

mp

176-179  °C

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4+,5+,6-,7-/m1/s1

InChI key

HOVAGTYPODGVJG-VOQCIKJUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A β-D-galactopyranoside having a methyl substituent at the anomeric position.

Biochem/physiol Actions

Methyl galactoside is a hexose involved in the metabolism of 2-deoxygalactose.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mayer et al.
Journal of the American Chemical Society, 123(25), 6108-6117 (2001-06-21)
A protocol based on saturation transfer difference (STD) NMR spectra was developed to characterize the binding interactions at an atom level, termed group epitope mapping (GEM). As an example we chose the well-studied system of galactose binding to the 120-kDa
NMR spectroscopy of hydroxyl protons in supercooled carbohydrates.
L Poppe et al.
Nature structural biology, 1(4), 215-216 (1994-04-01)
J W Dallinga et al.
Biological mass spectrometry, 23(12), 764-770 (1994-12-01)
The fast atom bombardment collision-induced dissociation mass spectra of the [M-H]- ions of the 2-, 3-, 4- and 6-deoxy derivatives from methyl beta-D-galactopyranoside and some related compounds have been recorded. The fragmentation reactions of these quasimolecular ions and of OD-labelled
Joel S Griffitts et al.
Science (New York, N.Y.), 307(5711), 922-925 (2005-02-12)
The development of pest resistance threatens the effectiveness of Bacillus thuringiensis (Bt) toxins used in transgenic and organic farming. Here, we demonstrate that (i) the major mechanism for Bt toxin resistance in Caenorhabditis elegans entails a loss of glycolipid carbohydrates;
Satoru Takahashi et al.
Epilepsy research, 80(1), 18-22 (2008-05-06)
Glucose transporter 1 (GLUT1) deficiency syndrome is caused by a deficit in glucose transport to the brain during the pre- and postnatal periods. Here, we report two cases of GLUT1 deficiency syndrome diagnosed on the basis of clinical features, reduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service