Skip to Content
Merck
All Photos(3)

Key Documents

L6004

Sigma-Aldrich

Lincomycin hydrochloride

≥90% (TLC)

Synonym(s):

Epilincomycin, Lincomycin HCL, Lincocin hydrochloride, Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-2-pyrrolidinecarboxamido)-1-thio-D-erythro-α-D-galactooctopyranoside hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34N2O6S · HCl
CAS Number:
Molecular Weight:
443.00
Beilstein:
4171650
EC Number:
MDL number:
UNSPSC Code:
51284507
PubChem Substance ID:
NACRES:
NA.85

Assay

≥90% (TLC)

potency

800-900 units per mg

color

white to faint yellow

solubility

H2O: soluble 50 mg/mL

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

InChI

1S/C18H34N2O6S.ClH/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H/t9-,10-,11+,12-,13+,14-,15-,16-,18-;/m1./s1

InChI key

POUMFISTNHIPTI-BOMBIWCESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Lincomycin hydrochloride is an antibiotic used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding It is produced by Streptomyces lincolnensis var. lincolnensis.

Biochem/physiol Actions

Mode of action: Lincomycin inhibits bacterial protein synthesis by forming cross-links within the peptidyl transferase loop region of the 23S rRNA.†

Antimicrobial spectrum: Lincomycin hydrochloride is effective against gram-positive bacteria.

Preparation Note

This product is soluble in water at 50 mg/mL, yielding a clear, colorless solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Malke et al.
Antimicrobial agents and chemotherapy, 19(1), 91-100 (1981-01-01)
The phenomenon of zonal resistance to lincomycin, which is characteristic of most clinical isolates with lincomycin resistance in Streptococcus pyogenes, has been studied. These strains grow within a defined concentration range of lincomycin (approximately 60 to 200 microgram/ml), or at
Neelam Keswani et al.
Journal of biochemistry, 148(1), 71-84 (2010-04-02)
The thermodynamics of interaction of neomycin and lincomycin with bovine serum albumin (BSA) and human serum albumin (HSA) has been studied using isothermal titration calorimetry (ITC), in combination with UV-visible, steady state and time resolved fluorescence spectroscopic measurements. Neomycin is
Charlotte M M Gommers et al.
Plant physiology, 185(1), 67-76 (2021-02-26)
When germinating in the light, Arabidopsis (Arabidopsis thaliana) seedlings undergo photomorphogenic development, characterized by short hypocotyls, greening, and expanded cotyledons. Stressed chloroplasts emit retrograde signals to the nucleus that induce developmental responses and repress photomorphogenesis. The nuclear targets of these
Ching-Chih Tseng et al.
Plant molecular biology, 82(4-5), 375-392 (2013-05-07)
RNA editing is one of the post-transcriptional processes that commonly occur in plant plastids and mitochondria. In Arabidopsis, 34 C-to-U RNA editing events, affecting transcripts of 18 plastid genes, have been identified. Here, we examined the editing and expression of
Kazunori Miyata et al.
Photosynthesis research, 113(1-3), 165-180 (2012-07-17)
When visible light is excess, the photosynthetic machinery is photoinhibited. The extent of net photoinhibition of photosystem II (PSII) is determined by a balance between the rate of photodamage to D1 and some other PSII proteins and the rate of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service