Skip to Content
Merck
All Photos(3)

Key Documents

L5751

Sigma-Aldrich

L-Lysine dihydrochloride

≥98% (HPLC), suitable for ligand binding assays

Synonym(s):

(S)-2,6-Diaminohexanoic acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4CH(NH2)COOH · 2HCl
CAS Number:
Molecular Weight:
219.11
Beilstein:
4539715
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Lysine dihydrochloride, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to faint yellow

mp

200-206 °C (lit.)

SMILES string

Cl[H].Cl[H].NCCCC[C@H](N)C(O)=O

InChI

1S/C6H14N2O2.2ClH/c7-4-2-1-3-5(8)6(9)10;;/h5H,1-4,7-8H2,(H,9,10);2*1H/t5-;;/m0../s1

InChI key

JBBURJFZIMRPCZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Lysine is an essential proteinogenic α amino acid used in a wide range of applications including as a supplement in cell culture media; a substrate of enzymes such as L-lysine oxidase (EC 1.4.3.14); a component of poly-lysine polymers; and a substrate for oxidation and glycation mechanism studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachery R Jarrell et al.
Aging, 12(13), 13555-13570 (2020-06-20)
Acylcarnitines transport fatty acids into mitochondria and are essential for β-oxidation and energy metabolism. Decreased mitochondrial activity results in increased plasma acylcarnitines, and increased acylcarnitines activate proinflammatory signaling and associate with age-related disease. Changes in acylcarnitines associated with healthy aging
Liying Yao et al.
PloS one, 11(2), e0149948-e0149948 (2016-02-24)
Myeloid-derived suppressor cells (MDSCs) are potent suppressors of T cell immunity in tumors and inflammatory diseases. They are identified by surface expression of CD11b+Gr1+ in mice, and CD11b+Gr1+ cells accumulate in the livers of obese mice. However, many myeloid cells
Gong Zhang et al.
Journal of agricultural and food chemistry, 59(22), 12037-12046 (2011-10-11)
A sensitive and reliable LC-(ESI)MS/MS method was developed and validated for the simultaneous analysis of five common advanced glycation endproducts (AGEs) after enzymatic digestion in raw and roasted almonds. AGEs included carboxymethyl-lysine (CML), carboxyethyl-lysine (CEL), pyralline (Pyr), argpyrimidine (Arg-p), and
Xiu-Min Chen et al.
Journal of agricultural and food chemistry, 59(20), 11294-11303 (2011-09-23)
We investigated the antioxidant and anti-inflammatory activities of both crude and ultrafiltrated Maillard reaction (MR) products (MRPs) derived from sugar-amino acid MR models, comprising fructose (Fru), glucose (Glu) or ribose (Rib) reacted with glycine (Gly) or lysine (Lys), respectively. Crude
Hideaki Endo et al.
Analytical and bioanalytical chemistry, 391(4), 1255-1261 (2008-02-01)
L-lysine (L-Lys) in living bodies is critical for metabolism; therefore, determination of its levels in food is important. Most enzymatic methods for L-Lys analysis are performed using L-lysine oxidase (LyOx), but commercially manufactured LyOx is generally not highly selective for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service