Skip to Content
Merck
All Photos(2)

Key Documents

L4762

Sigma-Aldrich

Loperamide hydrochloride

98-102% (USP), powder, Ca2+ channel blocker

Synonym(s):

4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H33ClN2O2 · HCl
CAS Number:
Molecular Weight:
513.50
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Loperamide hydrochloride,

originator

Johnson & Johnson

SMILES string

Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4

InChI

1S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34H,17-22H2,1-2H3;1H

InChI key

PGYPOBZJRVSMDS-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

Application

Loperamide hydrochloride (HCl) has been used in inflammatory pain experiments and to assess the functionality of proteins. 54

Biochem/physiol Actions

Loperamide hydrochloride (HCl) is a non-selective Ca2+ channel blocker. At nanomolar concentrations, it binds to μ-opioid receptors. Loperamide HCl does not cross the blood-brain barrier.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Slightly soluble in water and dilute acids, freely soluble in methanol and in chloroform, soluble in ethanol (95%).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Church et al.
Molecular pharmacology, 45(4), 747-757 (1994-04-01)
The effects of the antidiarrheal agent loperamide on high-voltage-activated (HVA) calcium channel activity and excitatory amino acid-evoked responses in two preparations of cultured hippocampal pyramidal neurons were examined. In rat hippocampal neurons loaded with the calcium-sensitive dye fura-2, rises in
Olivia Benguettat et al.
PLoS pathogens, 14(9), e1007279-e1007279 (2018-09-05)
The digestive tract is the first organ affected by the ingestion of foodborne bacteria. While commensal bacteria become resident, opportunistic or virulent bacteria are eliminated from the gut by the local innate immune system. Here we characterize a new mechanism
Raphaël Weibel et al.
PloS one, 8(9), e74706-e74706 (2013-09-27)
Opiates are powerful drugs to treat severe pain, and act via mu opioid receptors distributed throughout the nervous system. Their clinical use is hampered by centrally-mediated adverse effects, including nausea or respiratory depression. Here we used a genetic approach to
Min Guk Kim et al.
Journal of personalized medicine, 10(4) (2020-10-15)
Unbalanced dietary habits and the consumption of high protein and instant foods cause an increase in constipation. Here, we evaluated the effects of galacto-oligosaccharide (GOS) on a rat model of loperamide-induced constipation by measuring various biological markers and cecal microbiota.
Halil Kacmaz et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 33(2), e13994-e13994 (2020-10-02)
Gastrointestinal (GI) motility is a complex physiological process that is critical for normal GI function. Disruption of GI motility frequently occurs in GI diseases or as side effects of therapeutics. Whole gut transit measurements, like carmine red leading-edge transit, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service