Skip to Content
Merck
All Photos(1)

Key Documents

L4646

Sigma-Aldrich

Lipids Cholesterol Rich from adult bovine serum

liquid, sterile-filtered, BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Bovine Serum Lipids, Cholesterol Rich Lipids, Lipids from Bovine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

product line

BioReagent

form

liquid

composition

Cholesterol, 9.0-11.0 g/L
Protein, 15.0-25.0 g/L

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

impurities

Adventitious viral agents, none detected (9 CFR; 113.53)
IgG, none detected (RID)
Microbial content, none detected (USP XXII)
Mycoplasma, none detected (Barile method)
≤6.0 EU/mg cholesterol endotoxin

storage temp.

2-8°C

General description

Cholesterol is a lipid molecule, found integrated to the cell membrane.

Application

Lipids Cholesterol Rich from adult bovine serum has been used as one of the lipid component to generate erythroblasts ex vivo. It has also been used as a standard to determine muscle cholesterol level, spectrophotometrically.

Biochem/physiol Actions

Cholesterol significantly affects the property of phospholipid membrane such as fluidity and membrane protein function. It exhibits a condensing effect towards phospholipids. Cholesterol-phospholipid interaction influences physiological properties of a lipid bilayer. Cholesterol serves as a precursor of bile acids, steroid hormones and vitamin D. Uncontrolled level of plasma cholesterol might lead to the risk of cardiovascular disorder.

Other Notes

Aqueous lipoprotein solution

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Humanized culture medium for clinical expansion of human erythroblasts
Migliaccio G, et al.
Cell Transplantation, 19(4), 453-469 (2010)
Influence of dietary canola oil and palm oil blend and refrigerated storage on fatty acids, myofibrillar proteins, chemical composition, antioxidant profile and quality attributes of semimembranosus muscle in goats
Adeyemi KD, et al.
Journal of Animal Science and Biotechnology, 6(1), 51-51 (2015)
Dietary cholesterol: from physiology to cardiovascular risk
Lecerf JM and De Lorgeril M
The British Journal of Nutrition, 106(1), 6-14 (2011)
A molecular view of the cholesterol condensing effect in DOPC lipid bilayers
Alwarawrah M, et al.
The Journal of Physical Chemistry B, 114(22), 7516 -77523 (2010)
The condensing effect of cholesterol in lipid bilayers
Hung WC, et al.
Biophysical Journal, 92(11), 3960-3967 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service