Skip to Content
Merck
All Photos(2)

Key Documents

L0517

Sigma-Aldrich

Leukotriene B4

~100 μg/mL in ethanol, ≥97%

Synonym(s):

[5S,12R]-Dihydroxy-[6Z,8E,10E,14Z]-eicosatetraenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97%

form

liquid

concentration

~100 μg/mL in ethanol

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C[C@@H](O)\C=C\C=C\C=C/[C@@H](O)CCCC(O)=O

InChI

1S/C20H32O4/c1-2-3-4-5-6-9-13-18(21)14-10-7-8-11-15-19(22)16-12-17-20(23)24/h6-11,14-15,18-19,21-22H,2-5,12-13,16-17H2,1H3,(H,23,24)/b8-7+,9-6-,14-10+,15-11-/t18-,19-/m1/s1

InChI key

VNYSSYRCGWBHLG-AMOLWHMGSA-N

Gene Information

human ... LTB4R(1241)
rat ... Ltb4r(59264)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Proinflammatory agent. Stimulates c-Fos and c-Jun proto-oncogene transcription in human monocytes. Stimulates chemotaxis, aggregation, and degranulation of polymorphonuclear leukocytes.

Features and Benefits

This compound is featured on the Leukotriene Receptors and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Data sheet which accompanies each product includes special handling instructions. Except where noted, method for hydrolysis of methyl esters is also described.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nicolas Cenac et al.
Gastroenterology, 149(2), 433-444 (2015-04-26)
In mice, activation of the transient receptor potential cation channels (TRP) TRPV1, TRPV4, and TRPA1 causes visceral hypersensitivity. These receptors and their agonists might be involved in development of irritable bowel syndrome (IBS). We investigated whether polyunsaturated fatty acid (PUFA)
Leukotriene A4 hydrolase/aminopeptidase, the gatekeeper of chemotactic leukotriene B4 biosynthesis.
Jesper Z Haeggström
The Journal of biological chemistry, 279(49), 50639-50642 (2004-09-02)
H E Claesson et al.
International journal of immunopharmacology, 14(3), 441-449 (1992-04-01)
Leukotriene (LT) B4 is a biologically active molecule derived from arachidonic acid via the 5-lipoxygenase pathway. It mediates certain inflammatory and immunological reactions. The role of LTB4 in the immune system has been questioned since lymphocytes have been regarded to
L Iversen et al.
The Journal of investigative dermatology, 100(3), 293-298 (1993-03-01)
In the present study, keratinocytes were coincubated with human neutrophils to determine whether or not an increase in leukotriene B4 formation can occur. Human keratinocytes used were cultured in serum-free, low-calcium medium, whereas neutrophils were purified from heparinized venous blood.
Shyam Panthi et al.
Eye & contact lens, 45(3), 171-181 (2018-10-12)
Lipid mediators of inflammation are a group of signaling molecules produced by various cells under physiological conditions and modulate the inflammatory process during various pathologic conditions. Although eicosanoids and F2-isoprostanes are recognized lipid mediators of inflammation, there is no consensus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service