Skip to Content
Merck
All Photos(1)

Key Documents

K1884

Sigma-Aldrich

S-(+)-Ketamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16ClNO · HCl
CAS Number:
Molecular Weight:
274.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

powder

drug control

USDEA Schedule III; stupéfiant (France); regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain)

storage temp.

2-8°C

SMILES string

Cl.CN[C@@]1(CCCCC1=O)c2ccccc2Cl

InChI

1S/C13H16ClNO.ClH/c1-15-13(9-5-4-8-12(13)16)10-6-2-3-7-11(10)14;/h2-3,6-7,15H,4-5,8-9H2,1H3;1H/t13-;/m0./s1

InChI key

VCMGMSHEPQENPE-ZOWNYOTGSA-N

Biochem/physiol Actions

Selective NMDA glutamate receptor antagonist.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Chopra et al.
PloS one, 8(12), e81320-e81320 (2013-12-19)
To promote cancer research and to develop innovative therapies, refined pre-clinical mouse tumor models that mimic the actual disease in humans are of dire need. A number of neoplasms along the B cell lineage are commonly initiated by a translocation
Adam L Halberstadt et al.
Psychopharmacology, 233(7), 1215-1225 (2016-01-14)
Methoxetamine (MXE) is a ketamine analog sold online that has been subject to widespread abuse for its dissociative and hallucinogenic effects. Previous studies have shown that MXE has high affinity for the phencyclidine (PCP) binding site located within the channel
M Nishimura et al.
Neuroscience letters, 274(2), 131-134 (1999-12-20)
Ketamine is usually administered as a racemate, which is composed of the two isomers, S(+)-and R(-)-ketamine. Recently, we have shown that racemic ketamine at clinical relevant concentrations specifically inhibits the transporter proteins for norepinephrine, dopamine and serotonin heterologously expressed in
T Kawasaki et al.
Anesthesia and analgesia, 89(3), 665-669 (1999-09-04)
The production of proinflammatory cytokines, such as tumor necrosis factor (TNF) a, interleukin (IL)-6, and IL-8, increases in patients with sepsis; marked production causes organ failure and septic shock. We previously reported that ketamine suppressed lipopolysaccharide (LPS)-induced TNF-alpha production in
Martin Chopra et al.
PloS one, 8(9), e75737-e75737 (2013-10-08)
Multiple activities are ascribed to the cytokine tumor necrosis factor (TNF) in health and disease. In particular, TNF was shown to affect carcinogenesis in multiple ways. This cytokine acts via the activation of two cell surface receptors, TNFR1, which is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service