Skip to Content
Merck
All Photos(3)

Key Documents

I8132

Sigma-Aldrich

scyllo-Inositol

≥98%

Synonym(s):

1,3,5/2,4,6-Hexahydroxycyclohexane, DTLET

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

SMILES string

O[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3+,4+,5-,6-

InChI key

CDAISMWEOUEBRE-CDRYSYESSA-N

Other Notes

Naturally occurring isomer of myo-inositol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kevin A Dasilva et al.
Experimental neurology, 223(2), 311-321 (2009-09-12)
Structural insight into the conformational changes associated with aggregation and assembly of fibrils has provided a number of targets for therapeutic intervention. Solid-state NMR, hydrogen/deuterium exchange and mutagenesis strategies have been used to probe the secondary and tertiary structure of
Daniela Fenili et al.
Journal of molecular medicine (Berlin, Germany), 85(6), 603-611 (2007-02-07)
Inositol is a simple polyol with eight naturally occurring stereoisomers. myo-Inositol, D-chiro- and epi-inositol have been examined as potential therapeutic agents for various diseases, with favorable results, but treatment with scyllo-inositol has not been previously investigated. Our laboratory has shown
Paul H Yancey et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 133(3), 667-676 (2002-11-22)
Shallow-living marine invertebrates use free amino acids as cellular osmolytes, while most teleosts use almost no organic osmolytes. Recently we found unusual osmolyte compositions in deep-sea animals. Trimethylamine N-oxide (TMAO) increases with depth in muscles of some teleosts, skates, and
Aaron Y Lai et al.
Biochimica et biophysica acta, 1822(10), 1629-1637 (2012-07-18)
scyllo-Inositol (SI) is an endogenous inositol stereoisomer known to inhibit aggregation and fibril formation of the amyloid-beta peptide (Aβ). Human clinical trials using SI to treat Alzheimer disease (AD) patients have shown potential benefits. In light of the growing therapeutic
Matthew Townsend et al.
Annals of neurology, 60(6), 668-676 (2006-12-29)
Despite progress in defining a pathogenic role for amyloid beta protein (Abeta) in Alzheimer's disease, orally bioavailable compounds that prevent its effects on hippocampal synaptic plasticity and cognitive function have not yet emerged. A particularly attractive therapeutic strategy is to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service