Skip to Content
Merck
All Photos(1)

Key Documents

I6635

Sigma-Aldrich

Monoclonal Anti-Secretory Component (IgA) antibody produced in mouse

clone GA-1, ascites fluid

Synonym(s):

IgA Antibody, Secretory Component Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

GA-1, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

indirect ELISA: 1:10,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

The secretory component is a single chain glycoprotein that may occur freely or as component of the secretory immunoglobulin A (SIgA). IgA regulates the physiological balance between commensal intestinal bacteria and the defenses of the host immune system. It is the highest generated antibody isotype. IgAs are mainly secreted in the intestinal mucosa and prevent the commensal microorganisms from invading the intestines
Monoclonal Anti-Secretory Component (IgA) antibody is specific for secretory human IgA and the free secretory component. The antibody does not bind to human IgG, IgM and IgE.

Immunogen

Secretory component purified from human colostrum

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)
Monoclonal Anti-Secretory Component (IgA) antibody has been used in immunohistochemistry analysis and serological assays.
Monoclonal Anti-Secretory Component (IgA) antibody is suitable for use in EIA and western blot. The product may also be used for indirect ELISA (1:10,000).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muna Sapkota et al.
Alcoholism, clinical and experimental research, 41(12), 2093-2099 (2017-09-25)
Malondialdehyde (MDA) and acetaldehyde (AA) exist following ethanol metabolism and tobacco pyrolysis. As such, lungs of individuals with alcohol use disorders (AUDs) are a target for the effects of combined alcohol and cigarette smoke metabolites. MDA and AA form a
Min Zhao et al.
Experimental and therapeutic medicine, 14(5), 4033-4040 (2017-10-28)
Redox imbalance is established in various human diseases. Treatment of intestinal epithelial cells with hyperoxia for a prolonged period of time may cause serious effects on redox balance. Secretory component (SC) protein is secreted by intestinal epithelial cells, and has
B Simell et al.
The Journal of infectious diseases, 183(6), 887-896 (2001-03-10)
Local antibodies probably contribute to defense against Streptococcus pneumoniae. This study examined whether pneumococcal carriage and acute otitis media (AOM) induce mucosal antibodies to potential vaccine candidates pneumococcal surface adhesin A (PsaA), pneumolysin (Ply), and pneumococcal surface protein A (PspA).
Influence of reactive oxygen species on secretory component in the intestinal epithelium during hyperoxia
Zhao M, et al.
Experimental and Therapeutic Medicine, 14(5), 4033-4040 (2017)
Alpha Fardah Athiyyah et al.
Iranian journal of microbiology, 11(4), 300-304 (2019-11-14)
Probiotics and prebiotics are known to regulate immune responses. A synbiotic is a product that combines probiotics and prebiotics in a single dosage form. In this study, we attempt to present the effects of a multispecies synbiotic on intestinal mucosa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service