Skip to Content
Merck
All Photos(2)

Documents

I5907

Sigma-Aldrich

Pyrophosphatase, Inorganic from Escherichia coli

recombinant, expressed in E. coli, lyophilized powder, ≥90%, ≥800 units/mg protein

Synonym(s):

Inorganic Pyrophosphatase, Pyrophosphate phosphohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

Assay

≥90%

form

lyophilized powder

specific activity

≥800 units/mg protein

mol wt

hexamer subunit mol wt 20 kDa

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Pyrophosphatase from E coli (E-PPase) has a broader pH optimum and contains four divalent cations per subunit. It comprises 175 amino acids with additional aspartate residue in the active site cavity. Structurally E-PPase is a homohexamer with six identical 20 kDa subunits. Magnesium is a cofactor for E-PPase.

Application

Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme catalyzing the reaction PPi + H2O → 2Pi.
It plays an important role in protein, RNA, and DNA synthesis.
Pyrophosphatase, Inorganic from Escherichia coli has been used as a component of transcription buffer.
Pyrophosphatase, inorganic from Escherichia coli has been used in assay for conjugation of ubiquitin and ubiquitin-like proteins. It has been used for one-pot three-enzyme system for synthesis of Lewis x and sialyl Lewis x antigens.

Biochem/physiol Actions

Pyrophosphatase from E coli (E-PPase) is an essential enzyme in yeast and bacteria The active site residues are crucial for binding to magnesium.

Other Notes

A homohexameric protein containing 175 amino acid residues per subunit, its activity is Mg2+ dependent. It is a relatively thermostable protein.

Unit Definition

One unit will release 1.0 μmole of inorganic orthophosphate per minute at pH 9 at 25 °C.

Physical form

Lyophilized powder in Tris-buffered salts containing protease inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elodie Laine et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(25), 11277-11282 (2010-06-11)
Allostery plays a key role in the regulation of the activity and function of many biomolecules. And although many ligands act through allostery, no systematic use is made of it in drug design strategies. Here we describe a procedure for
Directed evolution of glycopeptides using mRNA display
Horiya S, et al.
Methods in Enzymology, 597, 83-141 (2017)
Structure and function analysis of Escherichia coli inorganic pyrophosphatase: is a hydroxide ion the key to catalysis?
Salminen, T, et al.
Biochemistry, 34(3), 782-791 (1995)
Rahul Sadavarte et al.
Biotechnology progress, 33(1), 133-139 (2016-11-01)
Heterologous overexpression of genes in Escherichia coli has made it possible to obtain high titers of recombinant proteins. However, this can result in the formation of aggregated protein particles known as 'inclusion bodies'. Protein sequestered as inclusion body is inactive
The structure of E. coli soluble inorganic pyrophosphatase at 2.7
Kankare J, et al.
Protein engineering, design & selection : PEDS, 7(7), 823-830 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service