Skip to Content
Merck
All Photos(2)

Key Documents

I1774

Sigma-Aldrich

Iturin A from Bacillus subtilis

≥95% (HPLC), suitable for microbiological culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

Product Name

Iturin A from Bacillus subtilis, ≥95% (HPLC)

biological source

Bacillus subtilis

Quality Level

Assay

≥95% (HPLC)

form

powder

technique(s)

microbiological culture: suitable

color

white to yellow

solubility

ethanol: soluble 9.80-10.20 mg/mL

antibiotic activity spectrum

fungi

Mode of action

cell membrane | interferes

storage temp.

2-8°C

Application

Iturin A was used in the study of Bacillomycin D (an iturin with antifungal activity against Aspergillus flavus).

Biochem/physiol Actions

IturinA exhibits strong antifungal activity against pathogenic yeast and fungi. It interacts with the cytoplasmic membrane of the target cell forming ion conducting pores. Its mode of action could be attributed to its interaction with sterols and phospholipids. The compound causes the release of exo-vesicles from human erythrocytes.

Other Notes

A family of lipopeptides characterized by a heptapeptide cyclized with a C13-C17 β-amino fatty acid. Composed mainly of C14-C15 β-amino acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mizumoto et al.
Applied microbiology and biotechnology, 75(6), 1267-1274 (2007-04-25)
Enhanced production of the antibiotic iturin A by Bacillus subtilis RB14-CS reached 4.4 g L(-1) in SM medium containing soybean meal and maltose, which was 16-fold and 2.2-fold higher than that in original and modified number 3S media, respectively. When
Katarzyna Czaczyk et al.
Polish journal of microbiology, 57(4), 313-319 (2008-01-01)
Cell surface hydrophobicity (CSH) is recognised as a important factor in microbial adhesion to solid surfaces. Growth conditions have been found to determine the synthesis of extracellular molecules by microorganisms. It has major consequences in modification of bacterial surface properties
Chie Niisawa et al.
The Journal of general and applied microbiology, 54(3), 149-158 (2008-07-26)
A composting product of marine animal resources has been used as a fertilizer and a soil conditioner in Japan. This compost was produced by a repeated fed-batch fermentation system with three successive aerobic bioreactors. Composting temperature reached about 75 degrees
Jun Yuan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(26), 2746-2750 (2011-08-30)
Iturin A, a powerful antifungal surfactant, is a kind of bacterial lipopeptide produced by Bacillus strains. This study addresses the use of an aqueous two-phase system (ATPS) using ethanol/ammonium sulfate to extract iturin A from Bacillus amyloliquefaciens NJN-6 fermentation broth
Minakshi Grover et al.
Current microbiology, 60(2), 99-106 (2009-09-25)
Bacillus subtilis strain RP24, isolated from rhizoplane of field grown pigeon pea, exhibited in vitro antagonism against a wide range of phytopathogenic fungi. An attempt was made to partially purify and characterize the diffusible antifungal metabolite/s produced by the strain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service