Skip to Content
Merck
All Photos(1)

Key Documents

HYSHP010

Sigma-Aldrich

HyStem®-HP Cell Culture Scaffold Kit

Trial Kit, For 2.5 mLof hydrogel scaffold solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Customizable, well characterized and consistent, Hystem-HP hydrogel cell culture scaffolds provide a complex, three dimensional environment in which cells are able to proliferate, much as they would in vivo.
  • Customizable - each distinct cell type in a multicellular organism has its own unique microenvironment. Hystem-HP offers the flexibility to tailor the environments of cultured cells to mimic their in vivo counterparts and optimize proliferation and differentiation.
  • Synthetic - As a synthetic matrix, Hystem-HP offers greater control of the cellular environment than ambiguous extracted ECM alternatives. All components are well characterized and consistently formulated. Hystem-HP contains Hyaluronic acid (synthetic), Extralink (synthetic), Gelin-S (porcine), Heparin and Water.
  • Biologically accurate - Rich in hyaluronic acid and denatured collagen, the Hystem-HP hydrogel closely mimics the complex three dimensional cellular environments found in life, providing an optimal environment for culturing your cells.
Hystem-HP enables ionic binding and slow release of growth factors through thiolated heparin; growth factors can be integrated via the matrix rather than cell culture medium. Hystem-HP also enables the attachment of a wide variety of cell types through Gelin-S (thiolated gelatin).

The Hystem-HP Trial Kit enables researchers to evaluate the Hystem-HP technology in their own hands and confirm that Hystem-HP is the right choice for their research.

With Hystem-HP, researchers can tailor the incorporation of ECM proteins and growth factors as well as the elasticity of the hydrogel and cell incorporation (encapsulations or top plating) to create the specific 3D environment required by their cells.

HyStem HP is recommended for in vivo (xenografts and animal models) experimentation.

Legal Information

Hystem is a registered trademark of BIOTIME, INC

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service