Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.47
UNSPSC Code:
41116121
form
solution
Quality Segment
shelf life
Expiry date on the label
IVD
for in vitro diagnostic use
technique(s)
microbe id | staining: suitable
application(s)
hematology
histology
storage temp.
room temp
Application
Yellow counterstain suitable for a wide range of histological staining procedures.
Other Notes
Tartrazine (0.25%) and acetic acid (0.25%)
Still not finding the right product?
Explore all of our products under Tartrazine Solution
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Magali Millecamps et al.
The spine journal : official journal of the North American Spine Society, 15(12), 2524-2537 (2015-09-04)
Chronic low back pain is debilitating and difficult to treat. Depending on the etiology, responses to treatments vary widely. Although chronic low back pain is frequently related to intervertebral disc degeneration, the relationship between disc degeneration severity and clinical symptoms
Seunghwan Lee et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 38(6), 1238-1247 (2019-12-10)
Low back pain (LBP) is a leading cause of global disability. Multiple anatomical, cellular, and molecular factors are implicated in LBP, including the degeneration of lumbar intervertebral discs (IVDs). We previously described a mouse model that displays behavioral symptoms of
Carmen La Porta et al.
Pain, 161(7), 1442-1458 (2020-03-07)
Low back pain (LBP) is a highly prevalent and disabling condition whose initiating factors are poorly understood. It is known that psychological and physical stress is associated with LBP but the causal relationship, mechanisms, and mediators have not been elucidated
Global Trade Item Number
| SKU | GTIN |
|---|---|
| HT3028-250ML | 04061838207630 |