Skip to Content
Merck
All Photos(2)

Key Documents

HT110232

Sigma-Aldrich

Eosin Y Solution, Aqueous

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47
Pricing and availability is not currently available.

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.5 % (w/v) in water

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Other Notes

Certified Eosin Y, 0.5% (w/v) in water. Not acidified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Hachmöller et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 44, 71-75 (2017-10-03)
The influence of rhodanine and haematoxylin and eosin (HE) staining on the copper distribution and concentration in liver needle biopsy samples originating from patients with Wilson's disease (WD), a rare autosomal recessive inherited disorder of the copper metabolism, is investigated.
Wei-Ting Chen et al.
Cell, 182(4), 976-991 (2020-07-24)
Although complex inflammatory-like alterations are observed around the amyloid plaques of Alzheimer's disease (AD), little is known about the molecular changes and cellular interactions that characterize this response. We investigate here, in an AD mouse model, the transcriptional changes occurring
Ivana Dinulovic et al.
Skeletal muscle, 6, 38-38 (2016-11-12)
Skeletal muscle tissue has an enormous regenerative capacity that is instrumental for a successful defense against muscle injury and wasting. The peroxisome proliferator-activated receptor γ coactivator 1α (PGC-1α) exerts therapeutic effects in several muscle pathologies, but its role in damage-induced
Marshall W Hogarth et al.
Nature communications, 8, 14143-14143 (2017-02-01)
Duchenne muscular dystrophy (DMD) is characterized by muscle degeneration and progressive weakness. There is considerable inter-patient variability in disease onset and progression, which can confound the results of clinical trials. Here we show that a common null polymorphism (R577X) in
Federica Francescangeli et al.
Journal of experimental & clinical cancer research : CR, 39(1), 2-2 (2020-01-09)
Quiescent/slow cycling cells have been identified in several tumors and correlated with therapy resistance. However, the features of chemoresistant populations and the molecular factors linking quiescence to chemoresistance are largely unknown. A population of chemoresistant quiescent/slow cycling cells was isolated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service