Skip to Content
Merck
All Photos(2)

Key Documents

HS2864

Arctic Squares® Freezer Rack for Upright Freezers

holds 2" freezer boxes, Holds 16 x boxes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122806
NACRES:
NB.22
Pricing and availability is not currently available.

material

steel

feature

Holds 16 x boxes

manufacturer/tradename

Heathrow Scientific HS2864

technique(s)

cryopreservation: suitable

size

139x571x238 mm

Features and Benefits

  • innovative horizontal drawer system for upright freezers
  • constructed of corrosion-resistant stainless steel with four drawers to hold 16 boxes
  • drawers feature a unique stop, that when fully extended pivot down allowing easy box access
  • steel drawer handles provide a user-friendly grip
  • freezer rack also includes a pull-out wire handle for easy removal off the freezer shelf
  • holds box height up to 53 mm
  • compatible with most standard 2" freezer storage boxes

Legal Information

Arctic Squares is a registered trademark of Heathrow Scientific LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hi, We are interested in this product but unfortunately is a little bit bigger for our freezers. I wonder if you have other racks similars to the model HS2864 but with this measures: overall L × W × H 13.9 x 55x 31.7 (55 or less). Thanks

    1 answer
    1. There are no freezer racks that meet close to or the exact specifications of L x W x H 13.9 cm, less than or equal to 55 cm, and 31.7 cm. The freezer racks offered are either too big or too small.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service