Skip to Content
Merck
All Photos(8)

Key Documents

HPA064845

Sigma-Aldrich

Anti-ACOX2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

Synonym(s):

BRCACOX, BRCOX, THCCox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VTVKGFTEALEKLENEPAIQQVLKRLCDLHAIHGILTNSGDFLHDAFLSGAQVDMARTAYLDLLRLIRKDAILLTDAFDFTDQCL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACOX2(8309)

General description

ACOX2 (acyl-CoA oxidase 2) is the rate limiting enzyme located on human chromosome 3p14. It codes for the peroxisomal branched-chain acyl-CoA oxidase. ACOX2 is expressed in the liver and kidney.

Immunogen

acyl-CoA oxidase 2, branched chain

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-ACOX2 antibody has been used in western blotting.

Biochem/physiol Actions

ACOX2 (acyl-CoA oxidase 2) participates in the β-oxidation of branched, long chain fatty acids and in the production of bile-acid precursor molecules. Loss of ACOX2 leads to inborn error of bile acid production with persistent hypertransaminasemia. ACOX2 may participates in the degradation of long branched fatty acids (phytanic and pristanic acids).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST90117

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ACOX2 deficiency: An inborn error of bile acid synthesis identified in an adolescent with persistent hypertransaminasemia.
Monte M J, et al.
Journal of Hepatology, 66(3), 581-588 (2017)
Expression of an estrogen-regulated variant transcript of the peroxisomal branched chain fatty acid oxidase ACOX2 in breast carcinomas.
Bj?rklund S S, et al.
BMC Cancer, 15(1), 524-524 (2015)
Marta Alonso-Peña et al.
Hepatology (Baltimore, Md.), 76(5), 1259-1274 (2022-04-09)
A variant (p.Arg225Trp) of peroxisomal acyl-CoA oxidase 2 (ACOX2), involved in bile acid (BA) side-chain shortening, has been associated with unexplained persistent hypertransaminasemia and accumulation of C27-BAs, mainly 3α,7α,12α-trihydroxy-5β-cholestanoic acid (THCA). We aimed to investigate the prevalence of ACOX2 deficiency-associated
Affinity proteomic profiling of plasma for proteins associated to area-based mammographic breast density.
Bystrom S, et al.
Breast Cancer Research, 20(1), 14-14 (2018)
ACOX2 deficiency: A disorder of bile acid synthesis with transaminase elevation, liver fibrosis, ataxia, and cognitive impairment.
Vilarinho S, et al.
Proceedings of the National Academy of Sciences of the USA, 113(40), 11289-11293 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service